Learn More
Thermo Scientific™ HyperSep™ Universal Vacuum Well Extraction Plate and Plugs
Product Code. 12732947 Shop All Thermo Scientific Products
Change view
Click to view available options
Type:
24 Position Extraction Plate Plugs
24 Well Extraction Plate
48 Position Extraction Plate Plugs
48 Well Extraction Plate
Unit Size:
Each
Pack of 24
Pack of 48
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Type unitSize
12732947 48 Well Extraction Plate Each
10321455 24 Position Extraction Plate Plugs Pack of 48
15277498 24 Well Extraction Plate Each
10219474 48 Position Extraction Plate Plugs Pack of 24
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Product Code. 12732947 Supplier Thermo Scientific™ Supplier No. 60104289
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Use these 24 or 48 well extraction plates and plugs with the Thermo Scientific™ HyperSep™ Universal Vacuum Manifold.
Use these 24 or 48 well extraction plates and plugs with Thermo Scientific™ HyperSep™ Universal Vacuum Manifold. This manifold process your samples in solid phase extraction SPE cartridges or 96 well plates.
TRUSTED_SUSTAINABILITY
Specifications
Description 48 Well Extraction Plate, 1 pk
For Use With (Application) SPE Sample Processing
For Use With (Equipment) Universal Vacuum Manifold
Product Line HyperSep
Type 48 Well Extraction Plate