Learn More
Thermo Scientific™ HyperSep™ Tip Microscale SPE Extraction Tips, 10-200μL
Product Code. 10775174 Shop All Thermo Scientific Products
Change view
Click to view available options
Sorbent:
BioBasic C18
BioBasic C4
BioBasic C8
HyperSep HILIC
HyperSep Hypercarb
HyperSep Hypercarb and C18 (Mix Mode)
HyperSep Titanium Dioxide
HyperSep Trypsin
HyperSep Zirconium Dioxide
Unit Size:
Pack of 96
9 product options available for selection
Product selection table with 9 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Sorbent Unit Size
10775174 HyperSep Hypercarb and C18 (Mix Mode) Pack of 96
10191213 BioBasic C18 Pack of 96
10566375 BioBasic C8 Pack of 96
10344083 BioBasic C4 Pack of 96
10465363 HyperSep HILIC Pack of 96
10055824 HyperSep Trypsin Pack of 96
10652094 HyperSep Titanium Dioxide Pack of 96
10516015 HyperSep Zirconium Dioxide Pack of 96
10223603 HyperSep Hypercarb Pack of 96
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 9 options available.
9 options
Product Code. 10775174 Supplier Thermo Scientific™ Supplier No. 60109213
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
missing translation for 'validMainframeMsg'
Get faster sample preparation with minimal sample loss with these revolutionary SPE micropipette tips.
Get faster sample preparation with minimal sample loss with these revolutionary SPE micropipette tips and achieve separation in volumes as low as 100nL.
Thermo Scientific™ HyperSep™ Microscale SPE Tips are available in many popular chemistries to suit a wide range of applications. They feature a unique micropipette tip that has the chromatographic material directly attached to its inner surface. No glues or polymers are used, so there's no contamination from the supporting matrix.
HyperSep Microscale Solid Phase Extraction Tips are ideal when you're working with small sample volumes. Typical applications include:
Mass spectrometry
Desalting
Protein purification
MALDI
Electrophoresis
HPCE
HPLC
CEC
Tips are available in 1-10µL capacity and 10-200µL capacity.
TRUSTED_SUSTAINABILITY
Specifications
Description C18 plus Hypercarb; 10 to 200 uL Volume;
For Use With (Application) Microscale Solid Phase Extraction
For Use With (Equipment) Micropipettes
Product Line HyperSep
Sorbent HyperSep Hypercarb and C18 (Mix Mode)
Type SPE Tips
Volume (Metric) 10 to 200 μL
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.