Learn More
Thermo Scientific™ HyperSep™ SPE Cartridges
Product Code. 30279934 Shop All Thermo Scientific Products
Change view
Click to view available options
Sorbent:
HLB (Reversed Phase Polymeric)
Mixed Mode Strong Anion Exchanger, Polymeric
Mixed Mode Strong Cation Exchanger, Polymeric
Type:
SPE Cartridge 10 mg/1 mL
SPE Cartridge 30 mg/1 mL
Unit Size:
Pack of 100
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Sorbent Type Unit Size
30279934 HLB (Reversed Phase Polymeric) SPE Cartridge 30 mg/1 mL Pack of 100
30279930 Mixed Mode Strong Cation Exchanger, Polymeric SPE Cartridge 30 mg/1 mL Pack of 100
30279931 Mixed Mode Strong Anion Exchanger, Polymeric SPE Cartridge 30 mg/1 mL Pack of 100
30279929 HLB (Reversed Phase Polymeric) SPE Cartridge 10 mg/1 mL Pack of 100
30279933 Mixed Mode Strong Cation Exchanger, Polymeric SPE Cartridge 10 mg/1 mL Pack of 100
30279932 Mixed Mode Strong Anion Exchanger, Polymeric SPE Cartridge 10 mg/1 mL Pack of 100
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Product Code. 30279934 Supplier Thermo Scientific™ Supplier No. 60107301HLB
Estimated Shipment: 20-11-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Achieve greater reproducibility with cleaner more consistent extracts compared to conventional SPE, phospholipid removal and protein precipitation products.
Grow your laboratory productivity. Thermo Scientific™ HyperSep™ SPE Cartridges revolutionize solid phase extraction (SPE). This first fritless SPE product range enables greater sample throughput in bioanalytical laboratories where low elution volumes combined with greater reproducibility, lower failure rates and cleaner extracts are key to achieving better productivity.
HyperSep products are available in a 10 mg/1 mL cartridge format. They provide highly reproducible, clean and consistent sample extracts for bioanalysis by utilizing the unique award winning fritless SPE technology. The enhanced performance of HyperSep cartridges gives you greater confidence in your analytical results and lowers operating costs - without compromising ease of use or requiring complex method development.
HyperSep cartridges provide unparalleled performance characteristics compared to conventional SPE, phospholipid removal and protein precipitation products.
What HyperSep Cartridges Mean to Your Analysis
  • Higher levels of reproducibility
  • Higher levels of extract cleanliness
  • Reduced solvent requirements
  • Confidence in results
TRUSTED_SUSTAINABILITY
Specifications
Bed Weight 30 mg
For Use With (Application) Bioanalytical Applications - Hydrophobic Retention of Neutral Compounds with Complementary Retention of Polar Analytes. An All-Purpose Phase.
Product Line HyperSep
Sorbent HLB (Reversed Phase Polymeric)
Type SPE Cartridge 30 mg/1 mL
Volume (Metric) 1 mL
Quantity 100/pk