Learn More
Thermo Scientific™ HyperSep™ Tip Microscale SPE Extraction Tips, 1-10μL
Product Code. 10703487 Shop All Thermo Scientific Products
Change view
Click to view available options
Sorbent:
BioBasic C18
BioBasic C4
BioBasic C8
HyperSep HILIC
HyperSep Hypercarb
HyperSep Hypercarb and C18 (Mix Mode)
HyperSep Titanium Dioxide
HyperSep Trypsin
HyperSep Zirconium Dioxide
Unit Size:
Pack of 96
9 product options available for selection
Product selection table with 9 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Sorbent Unit Size
10703487 BioBasic C8 Pack of 96
10600184 BioBasic C18 Pack of 96
10610184 BioBasic C4 Pack of 96
10456113 HyperSep Hypercarb Pack of 96
10098952 HyperSep Hypercarb and C18 (Mix Mode) Pack of 96
10223233 HyperSep HILIC Pack of 96
10426754 HyperSep Trypsin Pack of 96
10059002 HyperSep Titanium Dioxide Pack of 96
10556005 HyperSep Zirconium Dioxide Pack of 96
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 9 options available.
9 options
Product Code. 10703487 Supplier Thermo Scientific™ Supplier No. 60109202
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
missing translation for 'validMainframeMsg'
Get faster sample preparation with minimal sample loss with these revolutionary SPE micropipette tips.
Get faster sample preparation with minimal sample loss with these revolutionary SPE micropipette tips and achieve separation in volumes as low as 100nL.
Thermo Scientific™ HyperSep™ Microscale SPE Tips are available in many popular chemistries to suit a wide range of applications. They feature a unique micropipette tip that has the chromatographic material directly attached to its inner surface. No glues or polymers are used, so there's no contamination from the supporting matrix.
HyperSep Microscale Solid Phase Extraction Tips are ideal when you're working with small sample volumes. Typical applications include:
  • Mass spectrometry
  • Desalting
  • Protein purification
  • MALDI
  • Electrophoresis
  • HPCE
  • HPLC
  • CEC
Tips are available in 1-10μL capacity and 10-200μL capacity.
TRUSTED_SUSTAINABILITY
Specifications
Description 1 to 10 μL Volume
For Use With (Application) Microscale Solid Phase Extraction
For Use With (Equipment) Micropipettes
Particle Size 40 to 60 μm
Product Line HyperSep
Sorbent BioBasic C8
Type SPE Tips
Volume (Metric) 1 to 10 μL
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.