Learn More
Thermo Scientific™ HyperSep™ Javelin Direct-Connect Online SPE Columns
Shop All Thermo Scientific ProductsDescription
Clean-up and concentrate target analytes in minutes with Thermo Scientific™ HyperSep™ Online SPE Products. Available in several convenient formats, the columns utilize popular HyperSep SPE chemistries. When used with conventional HPLC systems, columns and appropriate switching valves, they provide:
- Effective removal of contaminants, such as proteins, from samples
- Fast, effective clean-up and concentration of target compounds
Thermo Scientific HyperSep Javelin Direct-Connect Online SPE Columns feature a unique fingertight design for convenience and efficiency.
HyperSep™ Retain™ PEP — for retention of polar and nonpolar analytes
HyperSep Retain-CX — for retention of basic and nonpolar analytes
HyperSep Retain-AX — for retention of acidic and nonpolar analytes
HyperSep™ Hypercarb™ — for retention of extremely polar analytes
Specifications
Specifications
| Description | HyperSep Retain-CX; 2.1 mm ID |
| For Use With (Application) | Online Solid Phase Extraction |
| For Use With (Equipment) | Conventional HPLC System |
| Product Line | HyperSep |
| Sorbent | HyperSep Retain CX |
| Type | Online Solid Phase Extraction Column |
| Diameter (Metric) Inner | 2.1 mm |