Learn More
Thermo Scientific™ HyperSep™ Glass Block Vacuum Manifold Collection Rack
Product Code. 10289454 Shop All Thermo Scientific Products
Change view
Click to view available options
Description:
Collection Rack for 16 Port Vacuum Manifold
Collection Rack for 24 Port Vacuum Manifold
Retaining Clips for Collection Rack
Unit Size:
Each
Pack of 12
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Description unitSize
10289454 Collection Rack for 24 Port Vacuum Manifold Each
10361245 Collection Rack for 16 Port Vacuum Manifold Each
10611226 Retaining Clips for Collection Rack Pack of 12
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 10289454 Supplier Thermo Scientific™ Supplier No. 60104252
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Select a 16- or 24-position collection rack for your Thermo Scientific™ HyperSep™ Glass Block Vacuum Manifold.
Select a 16- or 24-position collection rack for use with the Thermo Scientific™ HyperSep™ Glass Block Vacuum Manifold. The manifold enables solid phase extraction sample preparation using SPE cartridges. Extracts are collected into tubes in the collection rack.
HyperSep Glass Block Manifolds are durable, chemically resistant units designed to provide years of trouble-free extractions.
Polypropylene racks are highly resistant to chemical degradation and abuse, and are supplied according to the number of positions on the manifold lid
Vacuum control valve allows optimization of sample flow rates during extraction
Test tube racks allow the use of 12-13mm or 16mm disposable test tubes
Vacuum gauge and bleed valve enables easy monitoring of the vacuum process
Corner leg supports prevent damage to the luers and tips when the lid is not being used
HyperSep SPE Cartridges and Plates
HyperSep solid phase extraction cartridges offer reproducible, reliable sample preparation in a traditional format. Available in a wide range of chemistries and bed weights, the cartridges are ideal for large samples and conform to industry standard configurations, allowing efficient routine processing by vacuum, by positive pressure or centrifugation.
The HyperSep-96 system incorporates traditional solid phase extraction sorbents into 96-well plate format providing fast analysis without sacrificing accuracy or reproducibility. The innovative design of HyperSep-96 well plates provides flexibility for both method development and scale adjustment to 96-well without method revalidation. Well plates are customizable, allowing any arrangement of wells with differing sorbent chemistries for simultaneous processing. HyperSep-96 products are available in packs of 100 individual wells and in pre-assembled plates.
TRUSTED_SUSTAINABILITY
Specifications
Description Collection Rack for 24 Port Vacuum Manifold
For Use With (Application) Extract Collection
For Use With (Equipment) Glass Block Vacuum Manifold
Product Line HyperSep
Type Collection Rack
Quantity 1 Pk.