Learn More
Thermo Scientific™ HyperSep™ Filter Plates, 40¦L Bed Volume
Get simple, effective clean-up of proteins, peptides, DNA, RNA and other biomolecules in small-scale samples with these 96-well filter plates.
£207.00 - £270.00
Specifications
For Use With (Application) | Purification and Sample Preparation of Proteins, DNA, RNA and Other Biomolecules |
---|---|
Product Line | HyperSep |
Type | 96-Well Filter Plate |
Product Code | Brand | Description | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Description | Price | Quantity & Availability | |||||
10066254
|
Thermo Scientific™
60110-501 |
C18; 40 uL bed volume | |||||||
10240145
|
Thermo Scientific™
60110-502 |
C8; 40 uL bed volume | |||||||
10438104
|
Thermo Scientific™
60110-503 |
C4; 40 uL bed volume | |||||||
10782137
|
Thermo Scientific™
60110-504 |
Hypercarb; 40 uL bed volume | |||||||
10086454
|
Thermo Scientific™
60110-505 |
C18 plus Hypercarb; 40 uL bed volume | |||||||
10619095
|
Thermo Scientific™
60110-508 |
SCX; 40 uL bed volume | |||||||
10259744
|
Thermo Scientific™
60110-509 |
SAX; 40 uL bed volume | |||||||
Description
Achieve simple, effective purification and separation of proteins, peptides, DNA, RNA and other biomolecules in small-scale, microgram-level samples with Thermo Scientific™ HyperSep™ Filter Plates. Available in a range of chromatographic materials, the convenient 96-well plates can be used under vacuum.
These HyperSep Filter Plates are available with 40μL bed volumes.
HyperSep SPE Cartridges and PlatesHyperSep solid phase extraction cartridges offer reproducible, reliable sample preparation in a traditional format. Available in a wide range of chemistries and bed weights, the cartridges are ideal for large samples and conform to industry standard configurations, allowing efficient routine processing by vacuum, by positive pressure or centrifugation.
The HyperSep-96 system incorporates traditional solid phase extraction sorbents into 96-well plate format providing fast analysis without sacrificing accuracy or reproducibility. The innovative design of HyperSep-96 well plates provides flexibility for both method development and scale adjustment to 96-well without method revalidation. Well plates are customizable, allowing any arrangement of wells with differing sorbent chemistries for simultaneous processing. HyperSep-96 products are available in packs of 100 individual wells and in pre-assembled plates.
Specifications
Purification and Sample Preparation of Proteins, DNA, RNA and Other Biomolecules | |
HyperSep | |
96-Well Filter Plate |
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.