Learn More
- Home
- Products
- Solid Phase Extraction
- QuEChERS Dispersive SPE
- Thermo Scientific™ HyperSep™ Dispersive SPE Clean-up Products (QuEChERS)
Thermo Scientific™ HyperSep™ Dispersive SPE Clean-up Products (QuEChERS)
Product Code. 11534735 Shop All Thermo Scientific Products| Product Code. | Volume (Metric) | For Use With (Application) | Description | unitSize |
|---|---|---|---|---|
| 11534735 | 2 mL Centrifuge Tubes | QuEChERS Method | 150 mg MgSO4, 25 mg PSA | Pack of 10 |
| 11831282 | 2 mL Centrifuge Tubes | QuEChERS Method | 150 mg MgSO4, 50 mg PSA, 50 mg GCB | Pack of 100 |
| 11891062 | 2 mL Centrifuge Tubes | QuEChERS Method | 150 mg MgSO4, 50 mg PSA | Pack of 100 |
| 16883140 | 15 mL | QuEChERS Method | 900 mg anhydrous magnesium sulfate, 300 mg PSA | Pack of 50 |
| 10794817 | 2 mL Centrifuge Tubes | QuEChERS Method | 150 mg MgSO4, 50 mg PSA, 50 mg C18 | Pack of 100 |
| 16893140 | 15 mL | QuEChERS Method | 900 mg anhydrous magnesium sulfate, 150 mg PSA | Pack of 50 |
| 11851082 | 15 mL Centrifuge Tubes | QuEChERS Method | 900 mg MgSO4, 300 mg PSA, 150 mg GCB | Pack of 50 |
| 10769145 | 15 mL Centrifuge Tubes | QuEChERS Method | 900 mg MgSO4, 300 mg PSA, 150 mg C18 | Pack of 50 |
| 10639685 | 6 mL Centrifuge Tubes | Schenck Method | 400 mg PSA on Bottom, 200 mg GCB on Top, Separated by a Frit | Pack of 30 |
| 10289954 | 6 mL Centrifuge Tubes | Schenck Method | 500 mg PSA on Bottom, 250 mg GCB on Top, Separated by a Frit | Pack of 30 |
| 10127814 | 6 mL Centrifuge Tubes | Schenck Method | 500 mg PSA on Bottom, 500 mg GCB on Top, Separated by a Frit | Pack of 30 |
| 10650446 | 15 mL Centrifuge Tubes | QuEChERS Method | 900 mg MgSO4, 150 mg PSA | Pack of 50 |
| 10117364 | 15 mL Centrifuge Tubes | QuEChERS Method | 900 mg MgSO4, 150 mg PSA, 45 mg GCB | Pack of 50 |
| 10428664 | 15 mL Centrifuge Tubes | QuEChERS Method | 900 mg MgSO4, 150 mg PSA, 15 mg GCB | Pack of 50 |
| 10249644 | 2 mL Centrifuge Tubes | QuEChERS Method | 150 mg MgSO4, 25 mg PSA | Pack of 100 |
| 11524715 | 2 mL Centrifuge Tubes | QuEChERS Method | 150 mg MgSO4, 25 mg PSA, 25 mg C18 | Pack of 100 |
| 11534715 | 2 mL Centrifuge Tubes | QuEChERS Method | 150 mg MgSO4, 25 mg PSA, 2.5 mg GCB | Pack of 100 |
| 11544715 | 2 mL Centrifuge Tubes | QuEChERS Method | 150 mg MgSO4, 25 mg PSA, 7.5 mg GCB | Pack of 100 |
| 10320885 | 2 mL Centrifuge Tubes | QuEChERS Method | 150 mg MgSO4, 50 mg PSA, 50 mg C18, 50 mg GCB | Pack of 100 |
| 11564715 | 15 mL Centrifuge Tubes | QuEChERS Method | 1200 mg MgSO4, 400 mg PSA | Pack of 50 |
| 11574715 | 15 mL Centrifuge Tubes | QuEChERS Method | 1200 mg MgSO4, 400 mg PSA, 400 mg C18 | Pack of 50 |
| 10301995 | 15 mL Centrifuge Tubes | QuEChERS Method | 1200 mg MgSO4, 400 mg PSA, 400 mg C18, 400 mg GCB | Pack of 50 |
| 10066344 | 15 mL Centrifuge Tubes | QuEChERS Method | 900 mg MgSO4, 150 mg PSA, 150 mg C18 | Pack of 50 |
| 10649275 | 15 mL Centrifuge Tubes | QuEChERS Method | 150 mg MgSO4, 50 mg PSA, 50 mg GCB | Pack of 50 |
| 10497724 | 15 mL Centrifuge Tubes | QuEChERS Method | 150 mg MgSO4, 300 mg PSA, 150 mg Chlorofiltr | Pack of 50 |
| 15260359 | 2 mL Centrifuge Tubes | QuEChERS Method | 150 mg MgSO4, 50 mg PSA, 50 mg C18, 50 mg GCB | Pack of 10 |
| 11544735 | 2 mL Centrifuge Tubes | QuEChERS Method | 150 mg MgSO4, 25 mg PSA, 25 mg C18 | Pack of 10 |
| 11514735 | 15 mL Centrifuge Tubes | QuEChERS Method | 900 mg MgSO4, 150 mg PSA, 45 mg GCB | Pack of 10 |
| 11584725 | 15 mL Centrifuge Tubes | QuEChERS Method | 750 mg MgSO4, 250 mg PSA, 250 mg C18, 250 GCB | Pack of 10 |
| 11544725 | 6 mL Centrifuge Tubes | Schenck Method | 400 mg PSA on Bottom, 200 mg GCB on Top, Separated by a Frit | Pack of 10 |
| 11554735 | 2 mL Centrifuge Tubes | QuEChERS Method | 150 mg MgSO4, 25 mg PSA, 7.5 mg GCB | Pack of 10 |
| 11524725 | 2 mL Centrifuge Tubes | QuEChERS Method | 150 mg MgSO4, 50 mg PSA | Pack of 10 |
| 15207508 | 15 mL Centrifuge Tubes | QuEChERS Method | 900 mg MgSO4, 300 mg PSA, 150 mg C18 | Pack of 10 |
| 11564735 | 15 mL Centrifuge Tubes | QuEChERS Method | 900 mg MgSO4, 150 mg PSA, 150 mg C18 | Pack of 10 |
| 11524735 | 15 mL Centrifuge Tubes | QuEChERS Method | 900 mg MgSO4, 150 mg PSA, 15 mg GCB | Pack of 10 |
| 11534725 | 15 mL Centrifuge Tubes | QuEChERS Method | 900 mg MgSO4, 300 mg PSA, 150 mg GCB | Pack of 10 |
| 11594725 | 15 mL Centrifuge Tubes | QuEChERS Method | 900 mg MgSO4, 150 mg PSA | Pack of 10 |
| 15287498 | 15 mL Centrifuge Tubes | QuEChERS Method | 1200 mg MgSO4, 900 mg PSA | Pack of 50 |
| 15210369 | 15 mL Centrifuge Tubes | QuEChERS Method | 150 mg MgSO4, 300 mg PSA, 150 mg Chlorofiltr | Pack of 10 |
| 15220359 | 2 mL Centrifuge Tubes | QuEChERS Method | 150 mg MgSO4, 50 mg PSA, 50 mg GCB | Pack of 10 |
| 15270359 | 15 mL Centrifuge Tubes | QuEChERS Method | 1200 mg MgSO4, 400 mg PSA | Pack of 10 |
| 15240359 | 15 mL Centrifuge Tubes | QuEChERS Method | 900 mg MgSO4, 300 mg PSA | Pack of 10 |
| 15290349 | 2 mL Centrifuge Tubes | QuEChERS Method | 150 mg MgSO4, 50 mg PSA, 50 mg Chlorofiltr | Pack of 100 |
| 15210359 | 15 mL Centrifuge Tubes | QuEChERS Method | 150 mg MgSO4, 150 mg PSA, 50 mg C18 | Pack of 500 |
| 15220369 | 2 mL Centrifuge Tubes | QuEChERS Method | 150 mg MgSO4, 150 mg PSA | Pack of 100 |
| 15250359 | 2 mL Centrifuge Tubes | QuEChERS Method | 150 mg MgSO4, 25 mg PSA, 2.5 mg GCB | Pack of 10 |
| 15200369 | 15 mL Centrifuge Tubes | QuEChERS Method | 150 mg MgSO4, 50 mg PSA, 50 mg GCB | Pack of 10 |
| 15290359 | 15 mL Centrifuge Tubes | QuEChERS Method | 1200 mg MgSO4, 400 mg PSA, 400 mg C18, 400 mg GCB | Pack of 10 |
| 15280359 | 15 mL Centrifuge Tubes | QuEChERS Method | 1200 mg MgSO4, 400 mg PSA, 400 mg C18 | Pack of 10 |
| 15230359 | 2 mL Centrifuge Tubes | QuEChERS Method | 150 mg MgSO4, 50 mg PSA, 50 mg C18 | Pack of 10 |
| 10619875 | 15 mL Centrifuge Tubes | QuEChERS Method | 900 mg MgSO4, 300 mg PSA | Pack of 50 |
| 10259954 | 15 mL Centrifuge Tubes | QuEChERS Method | 750 mg MgSO4, 250 mg PSA, 250 mg C18, 250 GCB | Pack of 50 |
| 15297498 | 15 mL Centrifuge Tubes | QuEChERS Method | 4000 mg MgSO4, 1000 mg NaCl, 1000 mg C6H9Na3O9, 500 mg C6H6Na2O7 · 1.5 H2O | Pack of 50 |
| 15820645 | 2 mL Centrifuge Tubes | QuEChERS Method | 150 mg MgSO4, 50 mg PSA, 50 mg C18 | Pack of 100 |
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Description
Save time and achieve more accurate pesticide determinations using these pre-prepared dispersive SPE products for QuEChERS clean-up.
QuEChERS Dispersive SPE methods offer a convenient and effective approach for extraction and clean-up of pesticide residues in food and other complex matrices. The robust procedure offers a number of compelling advantages: high recoveries, accurate results, high sample throughput, low solvent and glassware usage, reduced labor and bench space and lower reagent costs.
Thermo Scientific™ HyperSep Dispersive SPE Clean-Up Products are available in a range of pre-prepared sorbent combinations, each containing the proper sorbents for optimum clean-up of analytes extracted using QuEChERS methods.
HyperSep Dispersive SPE Clean-Up Products contain PSA (primary/secondary amine) for the removal of organic acids and polar pigments among other compounds. Some products couple the PSA with endcapped C18 for the removal of most lipids and sterols, or graphitized carbon black for the removal of sterols and pigments such as chlorophyll.
(A variety of tube formats and bed weights are available to accommodate large and small sample sizes.)
All HyperSep Dispersive SPE products are pre-prepared and optimized for the method at hand, so you will:
- Save valuable preparation time
- Achieve more consistent analytical results due to consistency in products and rigorous quality control
- Get cleaner extracts and lower peak counts as a result of using highest quality, pre-prepared products
QuEChERS Technique
Developed by the USDA Eastern Regional Research Center in 2003, the QuEChERS (Quick, Easy, Cheap, Effective, Rugged and Safe) technique is increasingly becoming the technique of choice for extraction and clean-up of pesticide residues.
The QuEChERS method is a two-step process: extraction followed by clean-up. There are four variations of the QuEChERS method currently in use:
- The original QuEChERS method uses sodium chloride to enhance extraction
- Dispersive AOAC 2007.01 uses sodium acetate as a buffer, replacing sodium chloride
- The Dual-Phase variation introduces PSA and GCB to remove high levels of chlorophyll and plant sterols in the final extract without the loss of planar pesticides (polar aromatics) using an acetone:toluene solvent mix (3:1)
- European Version is similar to the AOAC method, except the extraction uses sodium chloride, sodium citrate dihydrate and disodium citrate sesquihydrate instead of sodium acetate
Specifications
Specifications
| Description | 150 mg MgSO4, 25 mg PSA |
| For Use With (Application) | QuEChERS Method |
| Product Line | HyperSep |
| Sorbent | Anhydrous Magnesium Sulfate, Primary/Secondary Amine |
| Type | QuEChERS Clean-up |
| Volume (Metric) | 2 mL Centrifuge Tubes |
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.