missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ HyperSep™ Dispersive SPE Clean-up Products (QuEChERS)

Product Code. 10301995 Shop All Thermo Scientific Products
Change view
Click to view available options
Volume (Metric):
15 mL
15 mL Centrifuge Tubes
2 mL Centrifuge Tubes
6 mL Centrifuge Tubes
For Use With (Application):
QuEChERS Method
Schenck Method
Description:
1200 mg MgSO4, 400 mg PSA
1200 mg MgSO4, 400 mg PSA, 400 mg C18
1200 mg MgSO4, 400 mg PSA, 400 mg C18, 400 mg GCB
1200 mg MgSO4, 900 mg PSA
150 mg MgSO4, 150 mg PSA
150 mg MgSO4, 150 mg PSA, 50 mg C18
150 mg MgSO4, 25 mg PSA
150 mg MgSO4, 25 mg PSA, 2.5 mg GCB
150 mg MgSO4, 25 mg PSA, 25 mg C18
150 mg MgSO4, 25 mg PSA, 7.5 mg GCB
150 mg MgSO4, 300 mg PSA, 150 mg Chlorofiltr
150 mg MgSO4, 50 mg PSA
150 mg MgSO4, 50 mg PSA, 50 mg C18
150 mg MgSO4, 50 mg PSA, 50 mg C18, 50 mg GCB
150 mg MgSO4, 50 mg PSA, 50 mg Chlorofiltr
150 mg MgSO4, 50 mg PSA, 50 mg GCB
400 mg PSA on Bottom, 200 mg GCB on Top, Separated by a Frit
4000 mg MgSO4, 1000 mg NaCl, 1000 mg C6H9Na3O9, 500 mg C6H6Na2O7 · 1.5 H2O
500 mg PSA on Bottom, 250 mg GCB on Top, Separated by a Frit
500 mg PSA on Bottom, 500 mg GCB on Top, Separated by a Frit
750 mg MgSO4, 250 mg PSA, 250 mg C18, 250 GCB
900 mg MgSO4, 150 mg PSA
900 mg MgSO4, 150 mg PSA, 15 mg GCB
900 mg MgSO4, 150 mg PSA, 150 mg C18
900 mg MgSO4, 150 mg PSA, 45 mg GCB
900 mg MgSO4, 300 mg PSA
900 mg MgSO4, 300 mg PSA, 150 mg C18
900 mg MgSO4, 300 mg PSA, 150 mg GCB
900 mg anhydrous magnesium sulfate, 150 mg PSA
900 mg anhydrous magnesium sulfate, 300 mg PSA
Unit Size:
Pack of 10
Pack of 100
Pack of 30
Pack of 50
Pack of 500
54 product options available for selection
Product selection table with 54 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Volume (Metric) For Use With (Application) Description unitSize
10301995 15 mL Centrifuge Tubes QuEChERS Method 1200 mg MgSO4, 400 mg PSA, 400 mg C18, 400 mg GCB Pack of 50
11831282 2 mL Centrifuge Tubes QuEChERS Method 150 mg MgSO4, 50 mg PSA, 50 mg GCB Pack of 100
11891062 2 mL Centrifuge Tubes QuEChERS Method 150 mg MgSO4, 50 mg PSA Pack of 100
16883140 15 mL QuEChERS Method 900 mg anhydrous magnesium sulfate, 300 mg PSA Pack of 50
10794817 2 mL Centrifuge Tubes QuEChERS Method 150 mg MgSO4, 50 mg PSA, 50 mg C18 Pack of 100
16893140 15 mL QuEChERS Method 900 mg anhydrous magnesium sulfate, 150 mg PSA Pack of 50
11851082 15 mL Centrifuge Tubes QuEChERS Method 900 mg MgSO4, 300 mg PSA, 150 mg GCB Pack of 50
10769145 15 mL Centrifuge Tubes QuEChERS Method 900 mg MgSO4, 300 mg PSA, 150 mg C18 Pack of 50
10639685 6 mL Centrifuge Tubes Schenck Method 400 mg PSA on Bottom, 200 mg GCB on Top, Separated by a Frit Pack of 30
10289954 6 mL Centrifuge Tubes Schenck Method 500 mg PSA on Bottom, 250 mg GCB on Top, Separated by a Frit Pack of 30
10127814 6 mL Centrifuge Tubes Schenck Method 500 mg PSA on Bottom, 500 mg GCB on Top, Separated by a Frit Pack of 30
10650446 15 mL Centrifuge Tubes QuEChERS Method 900 mg MgSO4, 150 mg PSA Pack of 50
10117364 15 mL Centrifuge Tubes QuEChERS Method 900 mg MgSO4, 150 mg PSA, 45 mg GCB Pack of 50
10428664 15 mL Centrifuge Tubes QuEChERS Method 900 mg MgSO4, 150 mg PSA, 15 mg GCB Pack of 50
10249644 2 mL Centrifuge Tubes QuEChERS Method 150 mg MgSO4, 25 mg PSA Pack of 100
11524715 2 mL Centrifuge Tubes QuEChERS Method 150 mg MgSO4, 25 mg PSA, 25 mg C18 Pack of 100
11534715 2 mL Centrifuge Tubes QuEChERS Method 150 mg MgSO4, 25 mg PSA, 2.5 mg GCB Pack of 100
11544715 2 mL Centrifuge Tubes QuEChERS Method 150 mg MgSO4, 25 mg PSA, 7.5 mg GCB Pack of 100
10320885 2 mL Centrifuge Tubes QuEChERS Method 150 mg MgSO4, 50 mg PSA, 50 mg C18, 50 mg GCB Pack of 100
11564715 15 mL Centrifuge Tubes QuEChERS Method 1200 mg MgSO4, 400 mg PSA Pack of 50
11574715 15 mL Centrifuge Tubes QuEChERS Method 1200 mg MgSO4, 400 mg PSA, 400 mg C18 Pack of 50
10066344 15 mL Centrifuge Tubes QuEChERS Method 900 mg MgSO4, 150 mg PSA, 150 mg C18 Pack of 50
10649275 15 mL Centrifuge Tubes QuEChERS Method 150 mg MgSO4, 50 mg PSA, 50 mg GCB Pack of 50
10497724 15 mL Centrifuge Tubes QuEChERS Method 150 mg MgSO4, 300 mg PSA, 150 mg Chlorofiltr Pack of 50
15260359 2 mL Centrifuge Tubes QuEChERS Method 150 mg MgSO4, 50 mg PSA, 50 mg C18, 50 mg GCB Pack of 10
11544735 2 mL Centrifuge Tubes QuEChERS Method 150 mg MgSO4, 25 mg PSA, 25 mg C18 Pack of 10
11514735 15 mL Centrifuge Tubes QuEChERS Method 900 mg MgSO4, 150 mg PSA, 45 mg GCB Pack of 10
11584725 15 mL Centrifuge Tubes QuEChERS Method 750 mg MgSO4, 250 mg PSA, 250 mg C18, 250 GCB Pack of 10
11544725 6 mL Centrifuge Tubes Schenck Method 400 mg PSA on Bottom, 200 mg GCB on Top, Separated by a Frit Pack of 10
11554735 2 mL Centrifuge Tubes QuEChERS Method 150 mg MgSO4, 25 mg PSA, 7.5 mg GCB Pack of 10
11524725 2 mL Centrifuge Tubes QuEChERS Method 150 mg MgSO4, 50 mg PSA Pack of 10
15207508 15 mL Centrifuge Tubes QuEChERS Method 900 mg MgSO4, 300 mg PSA, 150 mg C18 Pack of 10
11534735 2 mL Centrifuge Tubes QuEChERS Method 150 mg MgSO4, 25 mg PSA Pack of 10
11564735 15 mL Centrifuge Tubes QuEChERS Method 900 mg MgSO4, 150 mg PSA, 150 mg C18 Pack of 10
11524735 15 mL Centrifuge Tubes QuEChERS Method 900 mg MgSO4, 150 mg PSA, 15 mg GCB Pack of 10
11534725 15 mL Centrifuge Tubes QuEChERS Method 900 mg MgSO4, 300 mg PSA, 150 mg GCB Pack of 10
11594725 15 mL Centrifuge Tubes QuEChERS Method 900 mg MgSO4, 150 mg PSA Pack of 10
15287498 15 mL Centrifuge Tubes QuEChERS Method 1200 mg MgSO4, 900 mg PSA Pack of 50
15210369 15 mL Centrifuge Tubes QuEChERS Method 150 mg MgSO4, 300 mg PSA, 150 mg Chlorofiltr Pack of 10
15220359 2 mL Centrifuge Tubes QuEChERS Method 150 mg MgSO4, 50 mg PSA, 50 mg GCB Pack of 10
15270359 15 mL Centrifuge Tubes QuEChERS Method 1200 mg MgSO4, 400 mg PSA Pack of 10
15240359 15 mL Centrifuge Tubes QuEChERS Method 900 mg MgSO4, 300 mg PSA Pack of 10
15290349 2 mL Centrifuge Tubes QuEChERS Method 150 mg MgSO4, 50 mg PSA, 50 mg Chlorofiltr Pack of 100
15210359 15 mL Centrifuge Tubes QuEChERS Method 150 mg MgSO4, 150 mg PSA, 50 mg C18 Pack of 500
15220369 2 mL Centrifuge Tubes QuEChERS Method 150 mg MgSO4, 150 mg PSA Pack of 100
15250359 2 mL Centrifuge Tubes QuEChERS Method 150 mg MgSO4, 25 mg PSA, 2.5 mg GCB Pack of 10
15200369 15 mL Centrifuge Tubes QuEChERS Method 150 mg MgSO4, 50 mg PSA, 50 mg GCB Pack of 10
15290359 15 mL Centrifuge Tubes QuEChERS Method 1200 mg MgSO4, 400 mg PSA, 400 mg C18, 400 mg GCB Pack of 10
15280359 15 mL Centrifuge Tubes QuEChERS Method 1200 mg MgSO4, 400 mg PSA, 400 mg C18 Pack of 10
15230359 2 mL Centrifuge Tubes QuEChERS Method 150 mg MgSO4, 50 mg PSA, 50 mg C18 Pack of 10
10619875 15 mL Centrifuge Tubes QuEChERS Method 900 mg MgSO4, 300 mg PSA Pack of 50
10259954 15 mL Centrifuge Tubes QuEChERS Method 750 mg MgSO4, 250 mg PSA, 250 mg C18, 250 GCB Pack of 50
15297498 15 mL Centrifuge Tubes QuEChERS Method 4000 mg MgSO4, 1000 mg NaCl, 1000 mg C6H9Na3O9, 500 mg C6H6Na2O7 · 1.5 H2O Pack of 50
15820645 2 mL Centrifuge Tubes QuEChERS Method 150 mg MgSO4, 50 mg PSA, 50 mg C18 Pack of 100
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 54 options available.
54 options
This item is not returnable. View return policy
Product Code. 10301995 Supplier Thermo Scientific™ Supplier No. 60105226

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Save time and achieve more accurate pesticide determinations using these pre-prepared dispersive SPE products for QuEChERS clean-up.

QuEChERS Dispersive SPE methods offer a convenient and effective approach for extraction and clean-up of pesticide residues in food and other complex matrices. The robust procedure offers a number of compelling advantages: high recoveries, accurate results, high sample throughput, low solvent and glassware usage, reduced labor and bench space and lower reagent costs.

Thermo Scientific™ HyperSep Dispersive SPE Clean-Up Products are available in a range of pre-prepared sorbent combinations, each containing the proper sorbents for optimum clean-up of analytes extracted using QuEChERS methods.

HyperSep Dispersive SPE Clean-Up Products contain PSA (primary/secondary amine) for the removal of organic acids and polar pigments among other compounds. Some products couple the PSA with endcapped C18 for the removal of most lipids and sterols, or graphitized carbon black for the removal of sterols and pigments such as chlorophyll.

(A variety of tube formats and bed weights are available to accommodate large and small sample sizes.)

All HyperSep Dispersive SPE products are pre-prepared and optimized for the method at hand, so you will:

  • Save valuable preparation time
  • Achieve more consistent analytical results due to consistency in products and rigorous quality control
  • Get cleaner extracts and lower peak counts as a result of using highest quality, pre-prepared products

QuEChERS Technique

Developed by the USDA Eastern Regional Research Center in 2003, the QuEChERS (Quick, Easy, Cheap, Effective, Rugged and Safe) technique is increasingly becoming the technique of choice for extraction and clean-up of pesticide residues.

The QuEChERS method is a two-step process: extraction followed by clean-up. There are four variations of the QuEChERS method currently in use:

  • The original QuEChERS method uses sodium chloride to enhance extraction
  • Dispersive AOAC 2007.01 uses sodium acetate as a buffer, replacing sodium chloride
  • The Dual-Phase variation introduces PSA and GCB to remove high levels of chlorophyll and plant sterols in the final extract without the loss of planar pesticides (polar aromatics) using an acetone:toluene solvent mix (3:1)
  • European Version is similar to the AOAC method, except the extraction uses sodium chloride, sodium citrate dihydrate and disodium citrate sesquihydrate instead of sodium acetate
TRUSTED_SUSTAINABILITY

Specifications

Description 1200 mg MgSO4, 400 mg PSA, 400 mg C18, 400 mg GCB
For Use With (Application) QuEChERS Method
Product Line HyperSep
Sorbent Anhydrous Magnesium Sulfate, Primary/Secondary Amine, Endcapped C18, Graphitized Carbon Black
Type QuEChERS Clean-up
Volume (Metric) 15 mL Centrifuge Tubes

Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.