Learn More
Thermo Scientific™ HyperSep Carboxylic Acid (WCX) Fixed Well Plates
Weak cation exchange sorbent for extraction of strong bases
Brand:  Thermo Scientific™ 60307-332
Additional Details : Weight : 0.05000kg
Product Code. 15853250
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Description
Description
Applications:
  • Drugs and metabolites in biological fluids
  • Peptides in serum, plasma or biological fluids
  • Pesticides in water, food and beverages
  • Nucleic acids, nucleotides and surfactants
Specifications
Specifications
25 mg
HyperSep
1/pk