missing translation for 'onlineSavingsMsg'
Learn More

MedChemExpress Hypericin

Product Code. 16411418
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
Unit Size:
5mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16411418 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16411418 Supplier MedChemExpress Supplier No. HYN0453
 Restricted Item
Estimated Shipment: 07-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Hypericin is a naturally occurring substance found in Hypericum. Hypericin is an inhibitor of PKC (protein kinase C), MAO (monoaminoxidase), dopamine-beta-hydroxylase, reverse transcriptase, telomerase and CYP (cytochrome P450). Hypericin shows antitumor, antiviral, antidepressive activities, and can induce apoptosis.

Specifications

Content And Storage Powder -20°C 3 years, 4°C 2 years∣In solvent -80°C 6 months, -20°C 1 month
Molecular Weight (g/mol) 504.44 g/mol
Shipping Condition Room temperature
For Use With (Application) Cancer-Kinase/protease
CAS 548-04-9
Product Type Natural Products
Purity 98.0%
Solubility DMSO : 250 mg/mL (495.60 mM∣ultrasonic and warming and heat to 60°C)
Target Apoptosis∣Influenza Virus∣Antibiotic∣Monoamine Oxidase∣PKC∣Cytochrome P450∣Dopamine β-hydroxylase∣Reverse Transcriptase∣Telomerase
Quantity 5 mg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.