Learn More
Thermo Scientific™ HyPerforma™ Smartainer™ 3.0 Bioprocess Container (BPC), Aegis™ 5-14 film
Product Code. 31940834 Shop All Thermo Scientific Products
Change view
Click to view available options
Capacity (Metric):
200 L
500 L
Insert Type:
Horizontally-gusseted
Vertically-gusseted
Unit Size:
Each
Pack of 5
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity (Metric) Insert Type Unit Size
31940834 500 L Vertically-gusseted Each
17958221 200 L Horizontally-gusseted Pack of 5
17948221 500 L Horizontally-gusseted Pack of 5
31940830 500 L Horizontally-gusseted Each
17978261 500 L Vertically-gusseted Pack of 5
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Product Code. 31940834 Supplier Thermo Scientific™ Supplier No. SH31188.03
 Restricted Item
Estimated Shipment: 22-03-2027
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
The Thermo Scientific HyPerforma Smartainer 3.0 BPC provides end-users producing biologics or vaccine therapeutics with a large-volume, sterile liquid storage system for volumes ranging from 200 L to 3000 L—designed for the collection and storage of sterile process liquids such as cell culture media, process water or purification media. Options include temperature-control through integrated water jackets, BPC deploying hoists, as well as other options to optimize performance in your productions processes.
The Thermo Scientific HyPerforma Smartainer 3.0 BPC provides end-users producing biologics or vaccine therapeutics with a large-volume, sterile liquid storage system for volumes ranging from 200 L to 3000 L—designed for the collection and storage of sterile process liquids such as cell culture media, process water or purification media. Options include temperature-control through integrated water jackets, BPC deploying hoists, as well as other options to optimize performance in your productions processes.
TRUSTED_SUSTAINABILITY
Spezifikation
Capacity (Metric) 500 L
Product Type Bioprocess Container (BPC)
Insert Type Vertically-gusseted
Name des Produkts
Wählen Sie ein Thema
Indem Sie auf Absenden klicken, erklären Sie sich damit einverstanden, dass Fisher Scientific sich mit Ihnen in Verbindung setzen kann, um Ihr Feedback in diesem Formular zu bearbeiten. Wir werden Ihre Informationen nicht für andere Zwecke weitergeben. Alle bereitgestellten Kontaktinformationen werden in Übereinstimmung mit unserer Datenschutzrichtlinie aufbewahrt. Datenschutzrichtlinie.