Learn More
Description
Thermo Scientific™ HyPerforma™ 50 to 500L Tank Liners are available with or without bottom drain and are provided sterile for handling fluids in bioproduction applications.
Tank Liner Key Features:
- Available in sizes that fit into industry standard cylindrical tanks and in Thermo Scientific HyPerforma Drums (SV3011X, SV50076, and SV50517 series)
- Gusset design allows for a close fit to the tank
- Chamber constructed of Thermo Scientific CX3-9 film, which provides the same contact surface as other Thermo Scientific BPCs
- Supplied gamma irradiated
- Customization options are available by contacting one of our Thermo Scientific Representatives today
Standard Tank Liners for your liquid applications available in-stock in the following configurations:
SH30647.01 to .03:
- 50 to 200L Tank Liner
- Support Container Needed: Design to fit into HyPerforma Flat Bottom PE Drums (SV50076 series)
- Quantity: Each = 1 Liner
SH30647.04 to .07:
- 50 to 500L Tank Liner
- Support Container Needed: Designed to fit into Industry Standard Cylindrical Tanks (SV3011X series)
- Quantity: Each = 1 Liner
SH30646 Series:
- 50 to 200L Tank Liner with One Bottom Drain
- Bottom Lineset 1: 72in. (182.9cm) of tubing leading to a Quick Disconnect (MPX series) Insert (male) connector
- Support Container Needed: Designed to fit into HyPerforma Conical Bottom PE Drums (SV50517 series)
Ordering Information:
Supplied double polybagged and irradiated with Certificate of Analysis.
Recommended for:
- Hydrations of powdered media and buffers
- Pooling of non-sterile solutions and fluids
- Non-aseptic collection of waste liquids from separation and purification systems
To order, use the part numbers listed in the above table.
Specifications
Specifications
| Green Features | Fewer resources used and less waste |
| Capacity (Metric) | 50 L |
| Material | Plastic |
| Product Type | Tank Liner |
| Outer Packaging | Packages are placed in strong cardboard cartons with product and lot identification. |
| Protective Inner Packaging | Packed and sealed in two independent outer layers in the Class 10,000/ISO Class 7 area. |
| For Use With (Equipment) | SV30111 |
| Sterilization Method | Irradiated |
| Product Line | HyPerforma |
| Film Description | CX3-9 |
| Show More |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.