missing translation for 'onlineSavingsMsg'
Learn More

HYPE Antibody (3F5), Novus Biologicals™

Product Code. 18386259 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mg
Unit Size:
0.01mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18386259 0.1 mg 0.01mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18386259 Supplier Novus Biologicals Supplier No. H00011153M02

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

HYPE Monoclonal antibody specifically detects HYPE in Human samples. It is validated for Western Blot, ELISA, ELISA
TRUSTED_SUSTAINABILITY

Specifications

Antigen HYPE
Applications Western Blot, ELISA, Sandwich ELISA
Classification Monoclonal
Clone 3F5
Conjugate Unconjugated
Dilution Western Blot 1:500, ELISA, Sandwich ELISA
Formulation In 1x PBS, pH 7.4
Gene Accession No. AAH01342
Gene Alias AMPylator FICD, EC 2.7.7.n1, FIC domain containing, FIC domain-containing protein, fic S-phase protein cell division homolog, HIP-13, HIP13UNQ3041, huntingtin interacting protein 13, Huntingtin interacting protein E, huntingtin interactor protein E, Huntingtin yeast partner E, Huntingtin-interacting protein 13, Huntingtin-interacting protein E, HYPEadenosine monophosphate-protein transferase FICD, MGC5623
Host Species Mouse
Immunogen HYPE (AAH01342, 1 a.a. ~ 449 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. MAVTEPKWVSVWSRFLWVTLLSMVLGSLLALLLPLGAVEEQCLAVLKGLYLLRSKPDRAQHAATKCTSPSTELSITSRGATLLVAKTKASPAGKLEARAALNQALEMKRQGKREKAQKLFMHALKMDPDFVDALTEFGIFSEEDKDIIQADYLYTRALTISPYHEKALVNRDRTLPLVEEIDQRYFSIIDSKVKKVMSIPKGNSALRRVMEETYYHHIYHTVAIEGNTLTLSEIRHILETRYAVPGKSLEEQNEVIGMHAAMKYINTTLVSRIGSVTISDVLEIHRRVLGYVDPVEAGRFRTTQVLVGHHIPPHPQDVEKQMQEFVQWLNSEEAMNLHPVEFAALAHYKLVYIHPFIDGNGRTSRLLMNLILMQAGYPPITIRKEQRSDYYHVLEAANEGDVRPFIRFIAKCTETTLDTLLFATTEYSVALPEAQPNHSGFKETLPVKP
Purification Method IgG purified
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline Cell Cycle and Replication
Primary or Secondary Primary
Gene ID (Entrez) 11153
Target Species Human
Content And Storage Aliquot and store at -20°C or -80°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.