missing translation for 'onlineSavingsMsg'
Learn More

Hypaphorine, TRC

Product Code. 30392492
Change view
Click to view available options
Quantity:
0.5 mg
1 mg
5 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30392492 5 mg
30395917 1 mg
30391245 0.5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30392492 Supplier Toronto Research Chemicals Supplier No. TRCH9986385MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 487-58-1
Molecular Formula C14H18N2O2
Molecular Weight (g/mol) 246.31
Synonym 1H-Indole-3-ethanaminium, α-carboxy-N,N,N-trimethyl-, inner salt, (αS)- (9CI, ACI), Hypaphorine (6CI), (1-Carboxy-2-indol-3-ylethyl)trimethylammonium hydroxide, inner salt (7CI), 1H-Indole-3-ethanaminium, α-carboxy-N,N,N-trimethyl-, inner salt, (S)- (ZCI), Ammonium, (1-carboxy-2-indol-3-ylethyl)trimethyl-, hydroxide, inner salt (8CI), (+)-Hypaphorine, L-Hypaphorine, Glyyunnanenine, Lenticin, Lenticine, Tryptophan betaine, Tryptophan, trimethylbetaine
IUPAC Name (2S)-3-(1H-indol-3-yl)-2-(trimethylazaniumyl)propanoate
SMILES C[N+](C)(C)[C@@H](Cc1c[nH]c2ccccc12)C(=O)[O-]

Specifications

Quantity 5 mg
InChI Formula InChI=1S/C14H18N2O2/c1-16(2,3)13(14(17)18)8-10-9-15-12-7-5-4-6-11(10)12/h4-7,9,13,15H,8H2,1-3H3/t13-/m0/s1
Formula Weight 246.1368
Chemical Name or Material Hypaphorine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.