Learn More
Safety First Aid HypaClens Sterile Eye Wash Pods
Product Code. 31759384
Change view
Click to view available options
Sterility:
Sterile
Unit Size:
Pack of 25
Product Code. Sterility Unit Size
31759384 Sterile Pack of 25
1 options
Product Code. 31759384 Supplier Safety First Aid Supplier No. E401APK25
 Restricted Item
Estimated Shipment: 04-12-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
The eyewash pods gently wash the eye and provide moisture-replenishing relief for eyes that have been contaminated with particles or liquids. Their 0.9% sterile saline solution is ideal for eye irrigation.
  • HypaClens Sterile Eyewash Pods provide eye wash in a compact and easily portable size.
  • These 20 mL pods are ideal for inclusion in first aid kits where 250 mL or 500 mL bottles would be impractical.
  • They can be used to cleanse the eye of dust, dirt, small insects and other irritants.
  • For minor, non-emergency use, these pods are the perfect alternative to using a larger bottle, as eyewash bottles are single-use and cannot be resealed.
  • However, the pods are also ideal in the workplace as a portable, rapid-response item for responding to eye contamination at the scene of an incident.
Specifications
Product Type Eyewash Solution
Quantity 25 x 20 mL
Sterility Sterile