Learn More
Safety First Aid HypaClens Eye Wash Dispenser
Product Code. 31759421
Change view
Click to view available options
Color:
Clear
Unit Size:
Each
Product Code. Color Unit Size
31759421 Clear Each
1 options
Product Code. 31759421 Supplier Safety First Aid Supplier No. E498
Estimated Shipment: 06-11-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
The Eye Wash Dispenser is a wall-mounted dispenser and ensures that in the event of an emergency eye wash is easily located.
  • The dispenser features clear instructions for using eye wash pods on front of the cabinet, enabling ordinary people to provide help in case of an emergency.
  • The integrated mirror is ideal for self-administration of eye wash.
  • The HypaClens 20 mL Eye Wash Dispenser is ideal in any workplace environment, including workplaces with higher number of employees.
  • Refills can be purchased separately when required.
Specifications
Color Clear
Flow Rate 20 gpm (Shower), 8.0 gpm (Eye and Face Wash)
Material Plastic
Mounting Wall
Product Type Portable Eyewash and Shower Fixtures