Learn More
Safety First Aid HypaClens Emergency Eye Wash Station, Value (2 x 500 mL Bottles)
Product Code. 31759416
Change view
Click to view available options
Color:
Clear
Unit Size:
Each
Product Code. Color Unit Size
31759416 Clear Each
1 options
Product Code. 31759416 Supplier Safety First Aid Supplier No. E5201
Estimated Shipment: 06-11-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
The Eye Wash Station is a wall-mounted cardboard dispenser that enables you to provide a litre of HypaClens sterile emergency eyewash quickly and easily. Simply secure the dispenser to a wall by inserting two nails or screws through the provided slots.
  • It is ideal for short-term construction projects, especially where no clean running water is available.
  • HypaClens Sterile Eyewash is a single-use medical device intended for emergency eye irrigation.
  • As per HSE guidance, the eye should be washed out with either clean water or sterile fluid from a sealed container for at least 10 min. following contamination from chemicals or small particles.
  • The eyewash is designed as a substitute for clean running water in first aid situations and is only intended to be used once.
  • Re-use of the bottle would mean you can no longer guarantee the contents are sterile and face the risk of infection.
  • For this reason, the bottle is designed so it cannot be resealed.
  • After use, any excess fluid remaining should be discarded.
Specifications
Color Clear
Flow Rate 20 gpm (Shower), 8.0 gpm (Eye and Face Wash)
Material Plastic
Mounting Wall
Product Type Portable Eyewash and Shower Fixtures