Learn More
Safety First Aid HypaClean Automatic Hand Gel Dispenser
Product Code. 31759422
Change view
Click to view available options
No. of Compartments:
1
Unit Size:
Each
Product Code. No. of Compartments Unit Size
31759422 1 Each
1 options
Product Code. 31759422 Supplier Safety First Aid Supplier No. Q2174
  • ONLINE EXCLUSIVE PRICE
    £32.09
    £29.95 / Pack of 1
    Save £2.14
    6% Off
Estimated Shipment: 05-11-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
It is compatible with liquid sanitiser, hand gel, foam sanitiser and spray sanitiser. It is easy to fit to any wall by the screws or double- sided tape. The empty cartridge allows the dispenser to be filled with any sanitiser substance, no pouches are required.
  • It is ideal for public places including businesses, hospitals, restaurants, stations and airports and due to the automatic feature it prevents individuals from touching the dispenser.
  • Each output from the dispenser is approximately 0.5 mL to 0.6 mL if filled with hand gel.
  • The delay between each output of hand gel is approximately 3 sec.
  • Requires 4x C battery to allow the automatic feature to work, these are not included.
  • The dispenser can be used approximately 12000 to 13000 times when running on a new set of batteries.
  • Includes dispenser, key, screw and anchors, double sided tape and an instruction sheet.
  • Dispenser supplied empty - gel must be purchased separately.
Specifications
Product Type Hand Sanitizer Dispenser
Color White
Holds Hand Sanitizer
Material Plastic
Mounting Wall Mount
No. of Compartments 1
Product Style Top Loading