missing translation for 'onlineSavingsMsg'
Learn More

((HYNIC)-D-alanyl)-L-proline-boronic Acid TFA Salt, TRC

Product Code. 30395037
Change view
Click to view available options
Quantity:
0.5 mg
2.5 mg
5 mg
Unit Size:
0.5mg
2.5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30395037 2.5 mg 2.5mg
30394167 5 mg -
30397062 0.5 mg 0.5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30395037 Supplier Toronto Research Chemicals Supplier No. TRCH9722252.5MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

Molecular Formula C13 H20 B N5 O4 . x C2 H F3 O2
Molecular Weight (g/mol) 435.16
Synonym ((R)-1-((R)-2-(6-Hydrazinylnicotinamido)propanoyl)pyrrolidin-2-yl)boronic Acid TFA Salt
IUPAC Name [(2R)-1-[(2R)-2-[(6-hydrazinopyridine-3-carbonyl)amino]propanoyl]pyrrolidin-2-yl]boronic acid;2,2,2-trifluoroacetic acid
SMILES C[C@@H](NC(=O)c1ccc(NN)nc1)C(=O)N2CCC[C@H]2B(O)O.OC(=O)C(F)(F)F

Specifications

InChI Formula InChI=1S/C13H20BN5O4.C2HF3O2/c1-8(13(21)19-6-2-3-10(19)14(22)23)17-12(20)9-4-5-11(18-15)16-7-9;3-2(4,5)1(6)7/h4-5,7-8,10,22-23H,2-3,6,15H2,1H3,(H,16,18)(H,17,20);(H,6,7)/t8-,10+;/m1./s1
Quantity 2.5 mg
Formula Weight 435.1537
Shipping Condition Room Temperature
Chemical Name or Material ((HYNIC)-D-alanyl)-L-proline-boronic Acid TFA Salt
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.