missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Hymenialdisine Methanoate, TRC
Click to view available options
Quantity:
1 mg
2 mg
5 mg
Unit Size:
1mg
2mg
5mg
Specifications
Specifications
| Chemical Name or Material | Hymenialdisine Methanoate |
| Molecular Formula | C11H10BrN5O2 . x(CH4O) |
| Quantity | 2 mg |
| Synonym | (4Z)-4-(2-Amino-1,5-dihydro-5-oxo-4H-imidazol-4-ylidene)-2-bromo-4,5,6,7-tetrahydro-pyrrolo[2,3-c]azepin-8(1H)-one Methanoate, (Z)-Hymenialdisine Methanoate |
| InChI Formula | InChI=1S/C11H10BrN5O2.CH4O/c12-6-3-5-4(7-10(19)17-11(13)16-7)1-2-14-9(18)8(5)15-6;1-2/h3,15H,1-2H2,(H,14,18)(H3,13,16,17,19);2H,1H3/b7-4-; |
| SMILES | CO.NC1=NC(=O)\C(=C\2/CCNC(=O)c3[nH]c(Br)cc23)\N1 |
| Molecular Weight (g/mol) | 356.18 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction