Learn More
Invitrogen™ HYLS1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA596993
Description
Immunogen sequence: YPHVESNVPS ETVSEASQRL RKPVMKRKVL RRKPDGEVLV TDESIISESE SGTENDQDLW DLRQRLMNVQ FQEDKESSFD VSQKFNLPHE YQGISQDQLI CSLQREGMGS PAYEQDLIVA SRPKSFILPK LDQLSRNRGK TDRVARYFEY KRDWDSIRLP GEDHRKELRW GVREQMLCRA EPQSKPQHIY VPNNYLVPTE KKRSALRWGV RCDLANGVIP RKLPFPLSPS; Positive Samples: MCF-7; Cellular Location: Cytoplasm.
This gene encodes a protein localized to the cytoplasm. Mutations in this gene are associated with hydrolethalus syndrome. Multiple alternatively spliced variants, encoding the same protein, have been identified.
Specifications
| HYLS1 | |
| Polyclonal | |
| Unconjugated | |
| HYLS1 | |
| 3010015K02Rik; HLS; hydrolethalus syndrome 1; hydrolethalus syndrome protein 1; Hydrolethalus syndrome protein 1 homolog; Hyls1; HYLS1 centriolar and ciliogenesis associated; HYLS1, centriolar and ciliogenesis associated; hypothetical LOC538735 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 219844 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ELISA, Western Blot | |
| 1.53 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q96M11 | |
| HYLS1 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 70-299 of human HYLS1 (NP_6594511). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.