missing translation for 'onlineSavingsMsg'
Learn More

Hygromycin B, TRC

Product Code. 30133557
Change view
Click to view available options
Quantity:
100 mg
1 g
5 g
Unit Size:
100mg
1g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30133557 5 g 5g
30135890 1 g 1g
18073040 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30133557 Supplier Toronto Research Chemicals Supplier No. TRCH9976005G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 31282-04-9
Molecular Formula C20 H37 N3 O13
Molecular Weight (g/mol) 527.52
Synonym D-Streptamine, O-6-amino-6-deoxy-L-glycero-D-galacto-heptopyranosylidene-(1→2-3)-O-β-D-talopyranosyl-(1→5)-2-deoxy-N3-methyl-, Hygromycin B (7CI), O-6-Amino-6-deoxy-L-glycero-D-galacto-heptopyranosylidene-(1→2-3)-O-β-D-talopyranosyl-(1→5)-2-deoxy-N3-methyl-D-streptamine, Antihelmycin, Hygromix 2.4, Hygromix-8, Hygrovetin, Hygrovetine
IUPAC Name (3'R,3aS,4S,4'S,5'R,6R,6'R,7S,7aS)-4-[(1R,2S,3R,5S,6R)-3-amino-2,6-dihydroxy-5-(methylamino)cyclohexyl]oxy-6'-[(1S)-1-amino-2-hydroxyethyl]-6-(hydroxymethyl)spiro[4,6,7,7a-tetrahydro-3aH-[1,3]dioxolo[4,5-c]pyran-2,2'-oxane]-3',4',5',7-tetrol
SMILES CN[C@H]1C[C@@H](N)[C@H](O)[C@@H](O[C@@H]2O[C@H](CO)[C@H](O)[C@@H]3OC4(O[C@H]([C@@H](N)CO)[C@H](O)[C@H](O)[C@H]4O)O[C@H]23)[C@@H]1O

Specifications

Quantity 5 g
InChI Formula InChI=1S/C20H37N3O13/c1-23-7-2-5(21)9(26)15(10(7)27)33-19-17-16(11(28)8(4-25)32-19)35-20(36-17)18(31)13(30)12(29)14(34-20)6(22)3-24/h5-19,23-31H,2-4,21-22H2,1H3/t5-,6+,7+,8-,9+,10-,11+,12-,13+,14-,15-,16+,17+,18-,19+,20?/m1/s1
Formula Weight 527.2326
Chemical Name or Material Hygromycin B
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.