missing translation for 'onlineSavingsMsg'
Learn More

Melford Hygromycin B Powder

Product Code. 30018330
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30018330 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30018330 Supplier Melford Supplier No. H750201.0
 Restricted Item
Estimated Shipment: 26-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Hygromycin B is an antibiotic that is derived from Streptomyces hygroscopicus and has broad-spectrum activity against a wide range of bacteria, yeast, and higher eukaryotes.

Hygromycin B is an antibiotic that is derived from Streptomyces hygroscopicus and has broad-spectrum activity against a wide range of bacteria, yeast, and higher eukaryotes.

In molecular biology and genetic engineering, Hygromycin B is widely employed as a selective agent. It is commonly used in the selection of cells or organisms that have been successfully transformed with a hygromycin resistance gene. The resistance gene encodes an enzyme called hygromycin phosphotransferase, which phosphorylates and inactivates Hygromycin B, allowing only the transformed cells to survive and propagate.

Hygromycin B is also used in cell culture applications. It is effective in preventing the growth of bacterial and fungal contaminants in cell cultures, ensuring the purity of cell lines. It is often added to the growth media at appropriate concentrations to selectively eliminate non-transformed or contaminated cells.

Specifications

Chemical Name or Material Hygromycin B, Powder
CAS 31282-04-9
Quantity 1 g
Molecular Formula C20H37N3O13
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.