missing translation for 'onlineSavingsMsg'
Learn More

Hygromycin B, MedChemExpress

Product Code. p-200096814
Encompass_Preferred
Click to view available options
Quantity:
200 mg
500 mg
1 g
5 g
Unit Size:
1g
200mg
500mg
5g
This item is not returnable. View return policy

Product Code. 16462156

Brand: Medchem Express HYB0490

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Hygromycin B is an aminoglycoside antibiotic active against prokaryotic and eukaryotic cells.

Hygromycin B is an aminoglycoside antibiotic active against prokaryotic and eukaryotic cells.

Specifications

Chemical Name or Material Hygromycin B
CAS 31282-04-9
Color White
Purity Grade Notes Research
Molecular Formula C20H37N3O13
Quantity 500 mg
Synonym Hygrovetine
Solubility Information DMSO : 100 mg/mL (189.57 mM; ultrasonic and warming and heat to 60°C) ∣H2O : 50 mg/mL (94.78 mM; Need ultrasonic)
SMILES O[C@H]([C@H]1O)C2(O[C@@]([C@@H](N)CO)([H])[C@@H]1O)O[C@@]([C@@H](O[C@@H]3CO)O[C@@]([C@H]([C@H](N)C4)O)([H])[C@@H]([C@H]4NC)O)([H])[C@@]([C@H]3O)([H])O2
Molecular Weight (g/mol) 527.52
Formula Weight 527.52
Percent Purity 95.0%
Grade Research
Physical Form Powder
For Use With (Application) COVID-19-immunoregulation
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.