Learn More
Rotronic™ Hygrolog Datalogger
Product Code. 12899377
Product Code. 12899377 Supplier Rotronic™ Supplier No. HL20DSET
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
handheld temperature & humidity, digital display, integrated probe, software and data cable, 3 x AAA (1.5V) alkaline batteries 113mm x 32mm x 72mm (w x d x h) 0 to 100% rh, -10°C to +60°C Rotronic