Learn More
Technical Service Consultants™ Hygiene Sponge Sampling Kit
Product Code. 15354162
Change view
Click to view available options
Quantity:
200Pack
Unit Size:
Pack of 200
Product Code. Quantity unitSize
15354162 200Pack Pack of 200
1 options
Product Code. 15354162 Supplier Technical Service Consultants™ Supplier No. TS/15B.
Use Technical Service Consultants™ Hygiene Sponge Sampling Kit enables sampling in difficult to reach places, such as crevices and corners. It comes pre-dosed with a neutralizing buffer, swab heads, shaft materials,,tubes for convenient storage and handling and medical-grade plastics for assured quality. .
  • M40 Compliant
  • 2-year shelf life
  • Tamper-evident packaging
  • Swab heads and shaft materials
  • Rackable tubes
  • Medical grade plastics
Specifications
Product Type Hygiene Sponge Sampling Kits
Sterility Sterile
Includes Swab heads and shaft materials, rackable tubes, medical grade plastics
Quantity 200Pack
Storage Requirements 4 to 25°C. Store in dry and ventilated area; protect from light, heat and moisture in tightly closed original container
Tip Type Viscose