missing translation for 'onlineSavingsMsg'
Learn More

Ansell HyFlex™ 74-718 Cut-Resistant Gloves with Dyneema™ Diamond Technology

Product Code. 16885430
Change view
Click to view available options
Size:
7
8
9
10
11
Unit Size:
Pack of 12
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Size Unit Size
16885430 11 Pack of 12
16845430 7 Pack of 12
16855430 8 Pack of 12
16865430 9 Pack of 12
16875430 10 Pack of 12
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Product Code. 16885430 Supplier Ansell Supplier No. 74718110
Estimated Shipment: 08-02-2027
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Constructed with Dyneema™ Diamond Technology yarn for enhanced cut protection in high risk areas of the hand. Tuff Cuff™ II Technology helps gloves maintain their ergonomic shape during use, and after laundering.

  • Patent-pending engineered yarn provides EN 388 Cut Level 5 Protection
  • Dyneema™ Diamond Technology yarn enhances protection in high-risk areas of the hand
  • Incorporates Tuff Cuff™ II Technology to retain ergonomic shape during use and after laundering
  • Liner design helps reduce hand fatigue in high-stress areas of the hand by adjusting the knitting to match the shape of the hand, particularly in the knuckle areas
  • Advanced fibers help to control moisture and hand temperature in cold and warm environments
TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance (Europe) EN 420:2003 + A1:2009, REACH Compliant
Coating Material Uncoated
Color Blue
Cut Resistance Rating Level E
Hand Specific
Latex Free Yes
Material Nylon
Product Type Cut-resistant Gloves
Size 11
EN388 Standard 4X4XE
For Use With (Application) Food Processing
Glove Cuff Style Knit
Liner Material Dyneema™ Diamond Technology Yarn
Series 74-718
Yarn Gauge 10
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.