missing translation for 'onlineSavingsMsg'
Learn More

Ansell HyFlex™ 74-500 Food Safe Intercept Technology™ Gloves with 360° Protection

Product Code. p-7215002 Shop All Ansell Products
Click to view available options
Size:
7
8
9
10
Unit Size:
12 pairs

Product Code. 16445514

Brand: Ansell 74500070

Only null left
Add to basket
Add to basket

Intercept Technology™ provides Level F Cut Protection throughout the entire glove, even after repeat laundering. Extended cuff design for additional cut protection on the wrists.

  • Intercept™ is a specialized knitting technology that blends engineered, synthetic, and natural fibers into high performance yarns
  • Stain masking design for extended use life
  • Extended cuff expands cut protection to the wrists
  • Highly durable to retain dimensions and protection even after repeat laundering
  • Complies with FDA Food Handling Regulations 21 CFR 177.1520 and 21 CFR 177.2600
TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance (Europe) EN 420:2003 + A1:2009, EN 511:2006, REACH
Construction Knit
Coating Material Uncoated
Color Blue
Cut Resistance Rating Level F
Hand Ambidextrous
Latex Free Yes
Material Intercept Technology Yarn
Product Type Cut-resistant Gloves
Size 7
EN388 Standard 454XF
For Use With (Application) Food Processing
Gender Unisex
Glove Cuff Style Knit
Grip Design Smooth
Lining Lined
Packaging Type Case
Puncture Rating ANSI/ISEA 105-16 Level 4
Series 74-500
Thumb Style Wing
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.