missing translation for 'onlineSavingsMsg'
Learn More

Ansell HyFlex™ 72-400 Lightweight Cut-Resistant Spandex Gloves

Product Code. 16895420
Change view
Click to view available options
Size:
6
7
8
9
10
11
Unit Size:
Pack of 12
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Size unitSize
16895420 8 Pack of 12
16875420 6 Pack of 12
16885420 7 Pack of 12
16805430 9 Pack of 12
16815430 10 Pack of 12
16825430 11 Pack of 12
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
This item is not returnable. View return policy
Product Code. 16895420 Supplier Ansell Supplier No. 72400080
Add to basket
Add to basket
This item is not returnable. View return policy

Gloves provide a high level of resistance against cuts and abrasions without sacrificing dexterity. Lightweight design provides softness and flexibility so that gloves can be worn comfortably for an extended period of time.

  • Lightweight design provides softness and flexibility so that gloves can be comfortably worn for extended periods of time
  • Spandex elastic maintains a snug fit, even after laundering
  • Ambidextrous glove design extends useful life
  • Machine washable with non-caustic cleaners without affecting performance or durability
  • Compliant with FDA food handling requirements
TRUSTED_SUSTAINABILITY

Specifications

ANSI Abrasion Rating ANSI 3
Certifications/Compliance (Europe) EN 407:2004, EN 511:2006, EN 420:2003 + A1:2009, REACH Compliant
Coating Material Uncoated
Color Blue
Cut Resistance Rating ANSI A5
Hand Specific
Latex Free Yes
Material Intercept Technology Yarn
Product Type Mechanical Protection Gloves
Size 8
EN388 Standard 3541F
For Use With (Application) Food Processing
Glove Cuff Style Knit
Series 72-400
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.