missing translation for 'onlineSavingsMsg'
Learn More

Ansell ActivArmr™ 47-400 Medium Duty Nitrile Coated Oil Repellent Gloves

Product Code. 12640902 Shop All Ansell Products
Change view
Click to view available options
Size:
7
8
9
10
Unit Size:
12 pairs
Product Code. Size unitSize
12640902 7 12 pairs
12650902 8 12 pairs
12393619 9 12 pairs
12630902 10 12 pairs
4 options
This item is not returnable. View return policy

Product Code. 12640902

Brand: Ansell 47400070

Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

ActivArmr™ gloves combine the comfort of cotton or light leather gloves with the abrasion resistance and oil repellence of nitrile coated gloves.

  • ActivArmr™ gloves feature interlock knit lining, generous sizing, and contoured designs for improved comfort
  • Special nitrile formulation provides a combination of flexibility, mechanical resistance, and oil repellency
  • Natural hand shape is easier to wear for long periods
  • Gloves ensure cleanliness of metal sheets in the steel and motor industries
  • Silicone-free
TRUSTED_SUSTAINABILITY

Specifications

ANSI Abrasion Rating ANSI 4
Certifications/Compliance (Europe) EN 420:2003 + A1:2009; Category II
Construction Cut and Sewn
Coating Material Nitrile
Color Blue, Natural
Cut Resistance Rating ANSI A1
Exterior Finish No Texture
Hand Specific
Latex Free Yes
Material Cotton
Product Type Dip-coated Gloves
Size 7
For Use With (Application) Automotive, Machinery/Equipment, and Metal Fabrication
Glove Cuff Style Knit
Series 47-400
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.