Learn More
Ansell™ HyFlex 11-919 Gloves
Click to view available options
Size: 11; Color: White; Material: Nylon
Description
- Ideal glove for use in oily and grimy work environments.
- The seamless nylon liner provides an optimal fit, for comfort and dexterity.
- This glove is covered in a layer resistant to polluted pieces being handled.
Specifications
Coating Material | Nitrile |
Color | White, Blue |
Hand | Ambidextrous |
Material | Nylon |
Size | 11 |
Style | 11-919 |
Glove Cuff Style | Knit |
Length (Metric) | 220 to 260 mm |