missing translation for 'onlineSavingsMsg'
Learn More

Ansell HyFlex™ 11-917 Full Nitrile-Coated Seamless Knit Liquid Repellant Gloves

Product Code. 16079681 Shop All Ansell Products
Change view
Click to view available options
Size:
6
7
8
9
10
Unit Size:
12 pairs
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Size unitSize
16079681 6 12 pairs
16089681 7 12 pairs
16089891 8 12 pairs
16099891 9 12 pairs
16099681 10 12 pairs
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Product Code. 16079681 Supplier Ansell Supplier No. 11919060
Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

Gloves repel oil, grease, and dirt making them ideal for use in grimey work environments. Seamless nylon liner provides comfort, dexterity, and a secure fit.

  • Liquid repellent against oil, grease, and dirt
  • Outer layer is resistant to polluted objects
  • Seamless nylon liner
  • Suitable for food handling
TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance (Europe) EN 420:2003 + A1:2009, REACH Compliant
Coating Material Nitrile
Color Blue
Exterior Finish Rough
Hand Specific
Latex Free No
Material Nylon
Product Type Mechanical Protection Gloves
Size 6
EN388 Standard 4121A
For Use With (Application) Maintenance, General Handling, Quality Control, and Metal Fabrication
Glove Cuff Color White
Glove Cuff Style Knit
Series 11-919
Yarn Gauge 13
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.