Learn More
Ansell HyFlex™ 11-755 Cut Protection Medium Duty Task Gloves
Product Code. 18834417 Shop All Ansell Products
Change view
Click to view available options
Size:
6
11
Unit Size:
12 pairs
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Size Unit Size
18834417 11 12 pairs
18894407 6 12 pairs
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 18834417 Supplier Ansell Supplier No. 11755110
  • ONLINE EXCLUSIVE PRICE
    £181.10
    £173.82 / 12 pairs
    Save £7.28
    4% Off
View all promotions
In Stock
Add to basket
Edge
Add to basket
missing translation for 'validMainframeMsg'
Gloves provide reliable ANSI/ISEA 105-2024 A5/EN ISO E cut protection for medium-duty tasks.
  • Reliable protection: ANSI/ISEA 105-2024 A5/EN ISO E cut protection for everyday medium-duty tasks.
  • Flexible construction: Designed to enable comfortable, controlled handling and ease of movement.
  • Comfort and convenience: Touchscreen-compatible design supports comfortable all-day wear.
  • Confident grip: Polyurethane coating ensures secure handling in dry and light oil conditions.
  • Practical usability: Suitable for handling and assembly tasks requiring higher cut protection.
TRUSTED_SUSTAINABILITY
Specifications
Coating Material Polyurethane
Color Gray
Cut Resistance Rating Level E
Hand Specific
Latex Free Yes
Material Tungsten, HPPE, Polyester, Spandex, Polyurethane
Product Type Cut- and Abrasion-resistant Glove
Size 11