Learn More
Ansell HyFlex™ 11-754 - Ultralight Cut-Resistant Gloves
Product Code. 18060040
Change view
Click to view available options
Size:
6
7
8
9
10
11
12
Unit Size:
12 pairs
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Size unitSize
18060040 7 12 pairs
18030040 6 12 pairs
18040040 8 12 pairs
18050040 9 12 pairs
18000040 10 12 pairs
18020040 11 12 pairs
18070040 12 12 pairs
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
Product Code. 18060040 Supplier Ansell Supplier No. 11754070
Add to basket
Add to basket
Ultralight ANSI A4/EN ISO D-rated cut-resistant gloves with touchscreen compatibility.
  • Heightened defenses: Developed with INTERCEPT™ Cut Resistance Technology, and satisfying ANSI A4 and EN ISO D cut resistance standards, HyFlex 11-754 industrial gloves guard against lacerations and prevent common workplace injuries
  • Enhanced tactile sensitivity: Palm-dipped, with nylon liner, these PU-coated gloves benefit from advanced tactile sensitivity and touchscreen compatibility
  • Elevated comfort: Ultra-lightweight and fitted to natural hand contours, HyFlex 11-754 PPE gloves elicit minimal hand fatigue
  • Improved dexterity: These polyurethane gloves' ultralight design also increases flexibility and everyday ease of use
  • Certified skin-friendliness: HyFlex 11-754 industrial working gloves are Dermatest-approved for skin-friendliness
TRUSTED_SUSTAINABILITY
Specifications
Certifications/Compliance (Europe) Dermatologically Tested|Ecovadis label|EN 388:2016 +A1:2018|EN ISO 21420:2020
Coated Area Palm
Construction Knit
Coating Material Polyurethane
Color Gray
Cut Resistance Rating ANSI A4
Hand Ambidextrous
Latex Free No
Material Nylon, Spandex, Polyurethane
Product Type Cut- and Abrasion-resistant Glove
Size 7
EN388 Standard 3X41D
Gender Unisex
Glove Cuff Style Knit
Liner Material Nylon, Spandex, Stainless steel
Lining Lined
Thumb Style Wing
Yarn Gauge 18
Length (Metric) 225 to 285 cm