Learn More
Ansell HyFlex™ 11-754 - Ultralight Cut-Resistant Gloves
Product Code. 18070040
Change view
Click to view available options
Size:
6
7
8
9
10
11
12
Unit Size:
12 pairs
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Size Unit Size
18070040 12 12 pairs
18030040 6 12 pairs
18060040 7 12 pairs
18040040 8 12 pairs
18050040 9 12 pairs
18000040 10 12 pairs
18020040 11 12 pairs
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
Product Code. 18070040 Supplier Ansell Supplier No. 11754120
In Stock
Læg i indkøbskurv
Læg i indkøbskurv
missing translation for 'validMainframeMsg'
Denne vare kan ikke returneres. Se returpolicy
Ultralight ANSI A4/EN ISO D-rated cut-resistant gloves with touchscreen compatibility.
  • Heightened defenses: Developed with INTERCEPT™ Cut Resistance Technology, and satisfying ANSI A4 and EN ISO D cut resistance standards, HyFlex™ 11-754 industrial gloves guard against lacerations and prevent common workplace injuries
  • Enhanced tactile sensitivity: Palm-dipped, with nylon liner, these PU-coated gloves benefit from advanced tactile sensitivity and touchscreen compatibility
  • Elevated comfort: Ultra-lightweight and fitted to natural hand contours, HyFlex™ 11-754 PPE gloves elicit minimal hand fatigue
  • Improved dexterity: These polyurethane gloves' ultralight design also increases flexibility and everyday ease of use
  • Certified skin-friendliness: HyFlex™ 11-754 industrial working gloves are Dermatest™-approved for skin-friendliness
TRUSTED_SUSTAINABILITY
Tekniske data
Certifications/Compliance (Europe) Dermatologically Tested|Ecovadis label|EN 388:2016 +A1:2018|EN ISO 21420:2020
Coated Area Palm
Construction Knit
Coating Material Polyurethane
Color Gray
Cut Resistance Rating ANSI A4
Hand Ambidextrous
Latex Free No
Material Nylon, Spandex, Polyurethane
Product Type Cut- and Abrasion-resistant Glove
Size 12
EN388 Standard 3X41D
Gender Unisex
Glove Cuff Style Knit
Liner Material Nylon, Spandex, Stainless steel
Lining Lined
Thumb Style Wing
Yarn Gauge 18
Length (Metric) 225 to 285 cm
Vis mere Vis mindre
Produkttitel
Vælg et problem
Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.