missing translation for 'onlineSavingsMsg'
Learn More

Ansell HyFlex™ 11-725 Polyurethane-Coated Cut-Resistant Lycra™ Gloves

Product Code. 16689073 Shop All Ansell Products
Change view
Click to view available options
Size:
6
7
8
9
10
11
Unit Size:
12 pairs
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Size Unit Size
16689073 11 12 pairs
16639073 6 12 pairs
16649073 7 12 pairs
16659073 8 12 pairs
16669073 9 12 pairs
16679073 10 12 pairs
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Product Code. 16689073 Supplier Ansell Supplier No. 11725110
Outlet Corner
Use promo code 28080
View all promotions
In Stock
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

Gloves combine comfort and dexterity with a high degree of cut resistance for applications that involve sharp objects.

  • A balance of comfort, dexterity, and sure handling
  • High level of cut resistance for safe use in applications involving contact with sharp piece
  • Intercept™ is a specialized knitting technology that blends engineered, synthetic, and natural fibers into high performance yarns
  • Lycra™ in the liner/coating provides enhances dexterity and fit
  • Designed for use in dry, as well as slightly oily environments
TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance (Europe) EN 420:2003 + A1:2009, REACH Compliant
Coating Material Polyurethane
Color White
Cut Resistance Rating ANSI A3
Hand Specific
Latex Free No
Material Intercept Technology Yarn
Product Type Mechanical Protection Gloves
Size 11
EN388 Standard 4342B
For Use With (Application) Automotive Assembly, Construction, Maintenance, and Metal Fabrication
Glove Cuff Style Knit
Series 11-725
Yarn Gauge 15
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.