Learn More
Ansell Edmont™ HyFlex™ 11-630 Series Gray Polyurethane Medium Weight Gloves
Product Code. 11716663 Shop All Ansell Products
Change view
Click to view available options
Size:
6
7
8
9
10
11
Unit Size:
12 pairs
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Size unitSize
11716663 11 12 pairs
11706663 10 12 pairs
11726663 6 12 pairs
11736663 7 12 pairs
11746663 8 12 pairs
11756663 9 12 pairs
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Product Code. 11716663 Supplier Ansell Edmont™ Supplier No. 11630110
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Yarn structure contains an optimal rate of HPPE. Offers high levels of cut resistance for a safe use in controlling, handling and assembly applications involving contacts with sharp pieces.
  • The Lycra™ in the liners of the gloves as well as the polyurethane coating provides excellent flexibility and fit.
  • The specific formulation of the polyurethane dipping provides a secure grip on dry or slightly oily pieces.
  • The PU formulation also increases abrasion resistance on the gloves’ work surface.
  • With strong cut resistance
  • Knitted construction
TRUSTED_SUSTAINABILITY
Specifications
Back Material Polyurethane
Certifications/Compliance CE Mark, Standard CATEGORY II EN 388 : 4443
Cuff Material Polyurethane
Product Type Medium Weight Gloves
Size 11
Glove Cuff Style Knitwrist
Liner Material Dupont™ Kevlar™ aramid fibers
Lining Nylon/Lycra™/Dyneema™/Cotton/Dupont™ Kevlar™ aramid fiber
Palm Type Coated-palm
Quantity 144 Pairs
Resistant To High cut resistance