missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ansell Edmont™ HyFlex™ 11-625 Series White Polyurethane Medium Weight Gloves
Shop All Ansell Products
Click to view available options
Size:
6
7
8
9
10
Unit Size:
12 pairs
Descrizione
- Perfect balance of comfort and sure handling with superior cut resistance.
- The liners include an innovative yarn structure that contains an optimal rate of Dyneema™.
- Offers high levels of cut resistance
- The Lycra™ in the liners of the gloves as well as the polyurethane coating provides excellent flexibility and fit.
- Designed for use in dry, as well as slightly oily environments.
Specifica
Specifica
| Back Material | Polyurethane |
| Certifications/Compliance | CE Mark, Standard CATEGORY II EN 388 : 4342 |
| Cuff Material | Polyurethane |
| Product Type | Medium Weight Gloves |
| Size | 6 |
| Glove Cuff Style | Knitwrist |
| Liner Material | Dyneema™ |
| Lining | Spandex/Dyneema™ |
| Palm Type | Coated-palm |
| Quantity | 12 Pairs |
| Vedi altri risultati |
Correzione del contenuto del prodotto
Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.
Titolo del prodotto
Individuate un'opportunità di miglioramento?Condividi una correzione di contenuto