missing translation for 'onlineSavingsMsg'
Learn More

Ansell HyFlex™ 11-528 Cut-Resistant Polyester Spandex Gloves with Intercept Technology™

Product Code. 16009611 Shop All Ansell Products
Change view
Click to view available options
Size:
6
7
8
9
10
11
Unit Size:
12 pairs
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Size Unit Size
16009611 10 12 pairs
16069601 6 12 pairs
16079601 7 12 pairs
16089601 8 12 pairs
16099601 9 12 pairs
16019611 11 12 pairs
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
This item is not returnable. View return policy
Product Code. 16009611 Supplier Ansell Supplier No. 11528100
Estimated Shipment: 03-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Glass-free intercept liner is skin friendly, and provides ANSI A2 Cut Protection. NBR Foam coating enhances grip capabilities in dry, or light oil applications.

  • Glass-free intercept liner provides increased cut protection
  • Enhanced flexibility for range of motion and tactility when handling small parts
  • Grip performance for safe handling in dry to light oily applications
  • Intercept™ is a specialized knitting technology that blends engineered, synthetic, and natural fibers into high performance yarns
  • NBR Foam coating provides ANSI 4 Abrasion Resistance
  • DMF-free
TRUSTED_SUSTAINABILITY

Specifications

ANSI Abrasion Rating ANSI 4
Certifications/Compliance (Europe) EN 420:2003 + A1:2009, REACH Compliant
Coating Material Nitrile
Color Blue
Cut Resistance Rating ANSI A2
Hand Specific
Latex Free No
Material Intercept Technology Yarn
Product Type Mechanical Protection Gloves
Size 10
EN388 Standard 3X32B
For Use With (Application) Automotive Assembly, Metal Fabrication, Mining, and Transportation
Gender Unisex
Glove Cuff Style Knit
Series 11-528
Yarn Gauge 18
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.