Learn More
Ansell Edmont™ HyFlex™ 11-435 Series Polyurethane Gloves
Product Code. 11706643 Shop All Ansell Products
Change view
Click to view available options
Size:
6
7
8
9
10
11
Unit Size:
12 pairs
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Size unitSize
11706643 11 12 pairs
11796633 10 12 pairs
11716643 6 12 pairs
11726643 7 12 pairs
11736643 8 12 pairs
11746643 9 12 pairs
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Product Code. 11706643 Supplier Ansell Edmont™ Supplier No. 11435110
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
The first cut-resistant level 5 glove on the market made with water-based polyurethane to enhances the dexterity and comfort of the glove.
  • Ultra-strong fibers provide extreme resistance to cuts and burrs
  • Seamless plaited glove for very good fit, dexterity and flexibility
  • Excellent breathability and cool feeling
  • Gray palm coat hides soil and provides long wear, reducing overall replacement costs
  • Anti-static for improved safety in critical areas
  • Silicone-free
  • Oeko-Tex™ 100 approved
  • Ideal for the automotive and chemical industries, machinery and equipment and metal fabrication
TRUSTED_SUSTAINABILITY
Specifications
Back Material Polyurethane
Certifications/Compliance Standards CATEGORY II EN 388 : 4542, ANSI
Cuff Material Polyurethane
Product Type Medium Weight Gloves
Size 11
Glove Cuff Style Knitwrist
Liner Material Dyneema™
Lining Dyneema™
Palm Type Coated-palm
Quantity 144 Pairs
Resistant To Extreme resistance to cuts and burrs