missing translation for 'onlineSavingsMsg'
Learn More

Ansell™ Hyflex™ 11-100 ESD Control Gloves

Product Code. 15771277 Shop All Ansell Products
Click to view available options
Size:
8
9
Unit Size:
12 pairs
This item is not returnable. View return policy

Product Code. 15771277

Brand: Ansell™ 11100080

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Hyflex™ 11-100 ESD Control Gloves offer outstanding comfort and product protection from electrostatic discharge.

  • HyFlex™ 11-100 gloves are constructed with silver yarn for micro-electronic work or other applications where electrostatic discharge (ESD) must be avoided
  • The thin design improves the handling of even smaller components
  • Comfortable to wear
  • Nitrile foam palm coating ensures that surfaces stay clean and uncontaminated
  • Applications in the electronics, aerospace and manufacturing industries involving assembling, inspecting and packing small or delicate parts, or electrical component assembly and
    installation
TRUSTED_SUSTAINABILITY

Specifications

Coated Area Palm and front fingers, finger tips
Coating Material Nitrile Foam
Color Gray
Material Nylon
Product Type Hyflex 11-100 ESD Control Gloves
Size 8
For Use With (Application) Electronics and aviation component assembly or inspection
Glove Cuff Style Knitwrist
Palm Type Coated
Resistant To Electrostatic discharge (ESD)
Length (Metric) 220-275
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.