missing translation for 'onlineSavingsMsg'
Learn More

Hydroxyzine EP Impurity B (as Dihydrochloride), Mikromol™

Product Code. 30052632
Change view
Click to view available options
Quantity:
25 mg
Unit Size:
25mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30052632 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30052632 Supplier Mikromol Supplier No. MM0852.020025

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

Hydroxyzine EP Impurity B (as Dihydrochloride)
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 2-[2-[4-(Diphenylmethyl)piperazin-1-yl]ethoxy]ethanol Dihydrochloride (Decloxizine Dihydrochloride)
CAS 13073-96-6
Analyte or Component Names 2-[2-[4-(Diphenylmethyl)piperazin-1-yl]ethoxy]ethanol Dihydrochloride (Decloxizine Dihydrochloride)
Shipping Condition Room Temperature
Molecular Formula C21 H28 N2 O2 . 2 Cl H
Certifications/Compliance ISO 17025
Synonym Hydroxyzine Hydrochloride Imp. B (EP), Hydroxyzine Imp. B (EP), 2-[2-[4-(Diphenylmethyl)piperazin-1-yl]ethoxy]ethanol Dihydrochloride, Hydroxyzine Hydrochloride Imp. B (EP) as Dihydrochloride, Decloxizine Dihydrochloride, Hydroxyzine Hydrochloride Impurity B as Dihydrochloride, Hydroxyzine Impurity B as Dihydrochloride
InChI Formula InChI=1S/C21H28N2O2.2ClH/c24-16-18-25-17-15-22-11-13-23(14-12-22)21(19-7-3-1-4-8-19)20-9-5-2-6-10-20;;/h1-10,21,24H,11-18H2;2*1H
SMILES Cl.Cl.OCCOCCN1CCN(CC1)C(c2ccccc2)c3ccccc3
IUPAC Name 2-[2-(4-benzhydrylpiperazin-1-yl)ethoxy]ethanol;dihydrochloride
Molecular Weight (g/mol) 413.38
Quantity 25 mg
Formula Weight 412.1684 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.