missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Hydroxyzine Dihydrochloride, TRC
Specifications
Specifications
| Chemical Name or Material | Hydroxyzine Hydrochloride |
| CAS | 2192-20-3 |
| Molecular Formula | C21 H27 Cl N2 O2 . 2 Cl H |
| Quantity | 1 g |
| Synonym | Ethanol, 2-[2-[4-(p-chloro-α-phenylbenzyl)-1-piperazinyl]ethoxy]-, dihydrochloride (8CI), Ethanol, 2-[2-[4-[(4-chlorophenyl)phenylmethyl]-1-piperazinyl]ethoxy]-, dihydrochloride (9CI), 1-(p-Chlorobenzhydryl)-4-[2-(2-hydroxyethoxy)ethyl]diethylenediamine dihydrochloride, 2-[2-[4-(p-Chloro-α-phenylbenzyl)-1-piperazinyl]ethoxy]ethanol dihydrochloride, Alamon, Atarax, Atarax P Parenteral soln., Atarax dihydrochloride, Aterax, Atranine A, Durrax, Hydroxizine chloride, Hydroxyzine dihydrochloride, Hydroxyzine hydrochloride, Hydroxyzyne hydrochloride, Neurolax, Orgatrax, QYS, Quiess, Vistaril Parenteral, Vistaril Steraject |
| InChI Formula | InChI=1S/C21H27ClN2O2.2ClH/c22-20-8-6-19(7-9-20)21(18-4-2-1-3-5-18)24-12-10-23(11-13-24)14-16-26-17-15-25;;/h1-9,21,25H,10-17H2;2*1H |
| SMILES | Cl.Cl.OCCOCCN1CCN(CC1)C(c2ccccc2)c3ccc(Cl)cc3 |
| IUPAC Name | 2-[2-[4-[(4-chlorophenyl)-phenylmethyl]piperazin-1-yl]ethoxy]ethanol;dihydrochloride |
| Molecular Weight (g/mol) | 447.83 |
| Formula Weight | 446.1295 |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?