missing translation for 'onlineSavingsMsg'
Learn More

Hydroxyzine dihydrochloride, MedChemExpress

Product Code. 16402176
Encompass_Preferred
Click to view available options
Quantity:
100 mg
500 mg
Unit Size:
100mg
500mg
This item is not returnable. View return policy

Product Code. 16402176

Brand: Medchem Express HYB0548A

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Hydroxyzine dihydrochloride, a benzodiazepine antihistamine agent, acts as a orally active histamine H1-receptor and serotonin antagonist. Hydroxyzine dihydrochloride has anxiolytic effect and can be used forthe research of generalised anxiety disorder.

Hydroxyzine dihydrochloride, a benzodiazepine antihistamine agent, acts as a orally active histamine H1-receptor and serotonin antagonist. Hydroxyzine dihydrochloride has anxiolytic effect and can be used forthe research of generalised anxiety disorder.

Specifications

Chemical Name or Material Hydroxyzine dihydrochloride
Physical Form Solid
CAS 2192-20-3
Color White
Quantity 500 mg
Purity Grade Notes Research
Molecular Formula C21H29Cl3N2O2
For Use With (Application) COVID-19-immunoregulation
Solubility Information H2O : ≥ 150 mg/mL (334.95 mM)
SMILES ClC1=CC=C(C=C1)C(C2=CC=CC=C2)N3CCN(CCOCCO)CC3.Cl.Cl
Molecular Weight (g/mol) 447.83
Formula Weight 447.83
Percent Purity 98.05%
Grade Research
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.