missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Hydroxyzine Acetic Acid Hydrochloride Salt, TRC
Specifications
Specifications
| Chemical Name or Material | Hydroxyzine Acetic Acid Hydrochloride Salt |
| Analyte or Component Names | Hydroxyzine Acetic Acid Hydrochloride Salt |
| Shipping Condition | Room Temperature |
| Molecular Formula | C23 H29 Cl N2 O4 . xHCl |
| Synonym | 2-[2-[2-[4-[(4-chlorophenyl)phenylmethyl]-1-piperazinyl]ethoxy]ethoxy]-acetic acid, Acetic acid, [2-[2-[4-[(4-chlorophenyl)phenylmethyl]-1-piperazinyl]ethoxy]ethoxy]- (9CI), Cetirizine Dihydrochloride Imp. E (EP) |
| InChI Formula | InChI=1S/C23H29ClN2O4.ClH/c24-21-8-6-20(7-9-21)23(19-4-2-1-3-5-19)26-12-10-25(11-13-26)14-15-29-16-17-30-18-22(27)28;/h1-9,23H,10-18H2,(H,27,28);1H |
| Solution Type | Neat |
| SMILES | ClC1=CC=C(C=C1)C(N2CCN(CCOCCOCC(O)=O)CC2)C3=CC=CC=C3.Cl |
| IUPAC Name | 2-(2-(2-(4-((4-chlorophenyl)(phenyl)methyl)piperazin-1-yl)ethoxy)ethoxy)acetic acid hydrochloride |
| Industry Type | Pharmaceutical |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?