missing translation for 'onlineSavingsMsg'
Learn More

Hydroxyzine Acetic Acid Hydrochloride Salt, TRC

Product Code. 30392204
Change view
Click to view available options
Quantity:
2.5 mg
10 mg
25 mg
Unit Size:
10mg
2.50mg
25mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30392204 10 mg 10mg
30391368 2.5 mg 2.50mg
30405307 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30392204 Supplier Toronto Research Chemicals Supplier No. TRCH67115010MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Hydroxyzine Acetic Acid Hydrochloride Salt
Analyte or Component Names Hydroxyzine Acetic Acid Hydrochloride Salt
Shipping Condition Room Temperature
Molecular Formula C23 H29 Cl N2 O4 . xHCl
Synonym 2-[2-[2-[4-[(4-chlorophenyl)phenylmethyl]-1-piperazinyl]ethoxy]ethoxy]-acetic acid, Acetic acid, [2-[2-[4-[(4-chlorophenyl)phenylmethyl]-1-piperazinyl]ethoxy]ethoxy]- (9CI), Cetirizine Dihydrochloride Imp. E (EP)
InChI Formula InChI=1S/C23H29ClN2O4.ClH/c24-21-8-6-20(7-9-21)23(19-4-2-1-3-5-19)26-12-10-25(11-13-26)14-15-29-16-17-30-18-22(27)28;/h1-9,23H,10-18H2,(H,27,28);1H
Solution Type Neat
SMILES ClC1=CC=C(C=C1)C(N2CCN(CCOCCOCC(O)=O)CC2)C3=CC=CC=C3.Cl
IUPAC Name 2-(2-(2-(4-((4-chlorophenyl)(phenyl)methyl)piperazin-1-yl)ethoxy)ethoxy)acetic acid hydrochloride
Industry Type Pharmaceutical
Molecular Weight (g/mol) 469.4
Quantity 10 mg
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.