missing translation for 'onlineSavingsMsg'
Learn More

Hydroxyzine Acetic Acid Dihydrochloride, TRC

Product Code. 30003896
Change view
Click to view available options
Quantity:
25 mg
50 mg
100 mg
Unit Size:
100mg
25mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30003896 50 mg 50mg
30006014 25 mg 25mg
30012409 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30003896 Supplier Toronto Research Chemicals Supplier No. TRCH99700050MG
 Restricted Item
Estimated Shipment: 25-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 83881-56-5
Molecular Formula C23 H29 Cl N2 O4 . 2 Cl H
Molecular Weight (g/mol) 505.86
Synonym (RS)-2-[2-[2-[4-[(4-Chlorophenyl)phenylmethyl]piperazin-1-yl]ethoxy]ethoxy]-acetic Acid Dihydrochloride, Cetirizine Dihydrochloride Imp. E (EP) as Dihydrochloride, Ethoxycetirizine Dihydrochloride
IUPAC Name 2-[2-[2-[4-[(4-chlorophenyl)-phenylmethyl]piperazin-1-yl]ethoxy]ethoxy]acetic acid;dihydrochloride
SMILES Cl.Cl.OC(=O)COCCOCCN1CCN(CC1)C(c2ccccc2)c3ccc(Cl)cc3

Specifications

Quantity 50 mg
InChI Formula InChI=1S/C23H29ClN2O4.2ClH/c24-21-8-6-20(7-9-21)23(19-4-2-1-3-5-19)26-12-10-25(11-13-26)14-15-29-16-17-30-18-22(27)28;;/h1-9,23H,10-18H2,(H,27,28);2*1H
Formula Weight 504.1349
Percent Purity >95%
Chemical Name or Material (RS)-2-[2-[2-[4-[(4-Chlorophenyl)phenylmethyl]piperazin-1-yl]ethoxy]ethoxy]-acetic Acid Dihydrochloride (Ethoxycetirizine Dihydrochloride)
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.