missing translation for 'onlineSavingsMsg'
Learn More

MedChemExpress Hydroxytyrosol

Product Code. 16499018
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16499018 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16499018 Supplier MedChemExpress Supplier No. HYN0570
 Restricted Item
Estimated Shipment: 30-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Hydroxytyrosol (DOPET) is a phenolic compound with anti-oxidant, anti-atherogenic, anti-thrombotic, antimicrobial, anti-inflammatory and anti-tumour effects.

Specifications

Content And Storage Pure form -20°C 3 years, 4°C 2 years∣In solvent -80°C 6 months, -20°C 1 month
Molecular Weight (g/mol) 154.16 g/mol
Shipping Condition Room temperature
For Use With (Application) Cancer-programmed cell death
CAS 10597-60-1
Product Type Natural Products
Purity 99.6%
Solubility DMSO : 125 mg/mL (810.85 mM∣Need ultrasonic) ∣Ethanol : 50 mg/mL (324.34 mM∣Need ultrasonic)
Synonym DOPET∣ 3,4-Dihydroxyphenethyl alcohol∣ 3-Hydroxytyrosol
Target Endogenous Metabolite∣Bacterial∣Fungal
Quantity 10 mg
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.