missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Hydroxytacrine maleate salt, Thermo Scientific Chemicals
POA - £57.50
Chemical Identifiers
CAS | 118909-22-1 |
---|---|
Molecular Formula | C17H18N2O5 |
Molecular Weight (g/mol) | 330.34 |
MDL Number | MFCD00069287 |
InChI Key | NEEKVKZFYBQFGT-BTJKTKAUSA-N |
Synonym | velnacrine maleate, 9-amino-1,2,3,4-tetrahydroacridin-1-ol maleate, hydroxytacrine maleate, mentane, 1-hydroxy tacrine maleate, unii-wr9885gals, velnacrine maleate usan, wr9885gals, dsstox_cid_25158 |
PubChem CID | 5702293 |
IUPAC Name | 9-amino-1,2,3,4-tetrahydroacridin-1-ol;(Z)-but-2-enedioic acid |
SMILES | C1CC(C2=C(C3=CC=CC=C3N=C2C1)N)O.C(=CC(=O)O)C(=O)O |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15483409
|
Thermo Scientific Alfa Aesar
J60680.MD |
250 mg |
£57.50
250mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15473409
|
Thermo Scientific Alfa Aesar
J60680.03 |
1 g | |||||||
Description
Cholinesterase inhibitor. Potential Alzheimer's disease therapeutic
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
- Cholinesterase inhibitor
- Potential Alzheimer's disease therapeutic
Chemical Identifiers
118909-22-1 | |
330.34 | |
NEEKVKZFYBQFGT-BTJKTKAUSA-N | |
5702293 | |
C1CC(C2=C(C3=CC=CC=C3N=C2C1)N)O.C(=CC(=O)O)C(=O)O |
C17H18N2O5 | |
MFCD00069287 | |
velnacrine maleate, 9-amino-1,2,3,4-tetrahydroacridin-1-ol maleate, hydroxytacrine maleate, mentane, 1-hydroxy tacrine maleate, unii-wr9885gals, velnacrine maleate usan, wr9885gals, dsstox_cid_25158 | |
9-amino-1,2,3,4-tetrahydroacridin-1-ol;(Z)-but-2-enedioic acid |
Specifications
118909-22-1 | |
White | |
MFCD00069287 | |
NEEKVKZFYBQFGT-BTJKTKAUSA-N | |
9-amino-1,2,3,4-tetrahydroacridin-1-ol;(Z)-but-2-enedioic acid | |
5702293 | |
Solid |
171°C to 173°C | |
C17H18N2O5 | |
velnacrine maleate, 9-amino-1,2,3,4-tetrahydroacridin-1-ol maleate, hydroxytacrine maleate, mentane, 1-hydroxy tacrine maleate, unii-wr9885gals, velnacrine maleate usan, wr9885gals, dsstox_cid_25158 | |
C1CC(C2=C(C3=CC=CC=C3N=C2C1)N)O.C(=CC(=O)O)C(=O)O | |
330.34 | |
330.34 | |
Hydroxytacrine maleate salt |
Safety and Handling
missing translation for 'tsca' : No
Recommended Storage : Store at -20°C
RUO – Research Use Only